Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACUL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153810.4 | NP_722517.3 | 6 | 1357 | ||
GenBank mRNA | AJ544771.1 | - | 6 | 1141 | ||
AK097728.1 | - | 7 | 1077 | |||
AK131008.1 | - | 2 | 173 | |||
AK301182.1 | - | 4 | 384 | |||
AY743663.1 | - | 6 | 1357 | |||
BC038294.2 | - | 6 | 1357 | 4837841 |