Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LPAR1 SIRNA Assays ›
Gene Symbol:
LPAR1
Gene Name:
lysophosphatidic acid receptor 1
Gene Aliases:
EDG2, GPR26, Gpcr26, LPA1, Mrec1.3, VZG1, edg-2, rec.1.3, vzg-1
Chromosome Location:
Chr.9: 110873252 - 111039259 on Build GRCh38
UniGene ID:
Hs.126667
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
4050
Validated
Purification
In-Vivo Ready
Standard
HPLC
Size
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
Availability
Inventoried
Catalog #
AM51331
Price
6,301.00
Your Price
Online offer:
6301.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LPAR1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001401.3
NP_001392.2
3
1060
NM_057159.2
NP_476500.1
4
1138
XM_005251781.3
XP_005251838.1
4
1283
XM_005251782.3
XP_005251839.1
4
1377
XM_017014383.1
XP_016869872.1
4
1177
XM_017014384.1
XP_016869873.1
5
1551
XM_017014385.1
XP_016869874.1
6
1144
XM_017014386.1
XP_016869875.1
6
1444
XM_017014387.1
XP_016869876.1
6
1180
XM_017014388.1
XP_016869877.1
6
1363
XM_017014389.1
XP_016869878.1
5
1106
XM_017014390.1
XP_016869879.1
6
1141
XM_017014391.1
XP_016869880.1
6
1165
XM_017014392.1
XP_016869881.1
5
1408
XM_017014393.1
XP_016869882.1
6
1204
XM_017014394.1
XP_016869883.1
5
1333
XM_017014395.1
XP_016869884.1
5
1161
XM_017014396.1
XP_016869885.1
4
1103
XM_017014397.1
XP_016869886.1
5
1034
XM_017014398.1
XP_016869887.1
5
1277
XM_017014399.1
XP_016869888.1
5
1032
XM_017014400.1
XP_016869889.1
6
1393
XM_017014401.1
XP_016869890.1
5
1064
XM_017014402.1
XP_016869891.1
5
1121
XM_017014403.1
XP_016869892.1
6
1119
XM_017014404.1
XP_016869893.1
4
1317
XM_017014405.1
XP_016869894.1
6
1306
XM_017014406.1
XP_016869895.1
5
1040
XM_017014407.1
XP_016869896.1
5
1096
GenBank mRNA
AK022808.1
-
3
1060
AK290287.1
-
5
1413
AK296374.1
-
2
741
AK298590.1
-
2
717
AK299279.1
-
2
791
AY322546.1
-
2
712
BC030615.2
-
4
967
4815446
BC036034.1
-
4
1097
5272431
BC073167.1
-
5
1007
6193969
U78192.1
-
3
834
U80811.1
-
2
738
Y09479.1
-
3
834
Back To Top
More Information
Set Membership:
G-protein coupled receptor
Matching Taqman Gene Expression Assay:
Hs00173500_m1
Panther Classification:
Molecular Function -
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPK activity
G-protein coupled receptor signaling pathway
adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
activation of phospholipase C activity
positive regulation of cytosolic calcium ion concentration
regulation of cell shape
negative regulation of neuron projection development
oligodendrocyte development
cerebellum development
optic nerve development
corpus callosum development
negative regulation of cAMP biosynthetic process
bleb assembly
positive regulation of Rho protein signal transduction
myelination
positive regulation of apoptotic process
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of MAPK cascade
positive regulation of cytosolic calcium ion concentration involved in phospholipase C-activating G-protein coupled signaling pathway
positive regulation of stress fiber assembly
cell chemotaxis
positive regulation of dendritic spine development
cellular response to oxygen levels
positive regulation of smooth muscle cell chemotaxis
cellular response to 1-oleoyl-sn-glycerol 3-phosphate
G-protein alpha-subunit binding
G-protein coupled receptor activity
protein binding
PDZ domain binding
lysophosphatidic acid binding
lysophosphatidic acid receptor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0