Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASSF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206957.1 | NP_001193886.1 | 5 | 1436 | ||
NM_007182.4 | NP_009113.3 | 6 | 1543 | |||
NM_170712.2 | NP_733830.1 | 6 | 1253 | |||
NM_170713.2 | NP_733831.1 | 5 | 1347 | |||
NM_170714.1 | NP_733832.1 | 6 | 1555 | |||
XM_011533315.1 | XP_011531617.1 | 7 | 1632 | |||
XM_011533316.2 | XP_011531618.1 | 5 | 1451 | |||
GenBank mRNA | AF040703.2 | - | 5 | 1260 | ||
AF061836.1 | - | 5 | 1343 | |||
AF102770.2 | - | 6 | 1448 | |||
AF102771.2 | - | 6 | 1460 | |||
AF102772.2 | - | 6 | 1460 | |||
AF132675.1 | - | 6 | 1450 | |||
AF132676.1 | - | 5 | 1213 | |||
AF132677.1 | - | 6 | 1240 | |||
AF286217.1 | - | 5 | 1341 | |||
AF291719.1 | - | 4 | 1236 | |||
AK292638.1 | - | 5 | 1334 | |||
BC110412.1 | - | 6 | 1253 | 5756246 |