Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFNLR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170743.3 | NP_734464.1 | 7 | 4244 | ||
NM_173064.2 | NP_775087.1 | 7 | 4157 | |||
NM_173065.2 | NP_775088.1 | 6 | 4113 | |||
XM_017000479.1 | XP_016855968.1 | 6 | 4273 | |||
GenBank mRNA | AF439325.1 | - | 7 | 4217 | ||
AK160364.1 | - | 7 | 4218 | |||
BC140872.1 | - | 7 | 4292 | 9021740 |