Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSENEN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281532.1 | NP_001268461.1 | 4 | 535 | ||
NM_172341.2 | NP_758844.1 | 4 | 573 | |||
GenBank mRNA | AF220053.1 | - | 4 | 551 | ||
BC009575.1 | - | 4 | 493 | 4096271 | ||
BE563017.1 | - | 3 | 378 |