Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173479.3 | NP_775750.3 | 1 | 317 | ||
NR_135565.1 | 1 | 317 | ||||
XM_011526449.2 | XP_011524751.1 | 1 | 311 | |||
XM_011526450.2 | XP_011524752.1 | 1 | 311 | |||
XM_017026282.1 | XP_016881771.1 | 1 | 311 | |||
XM_017026283.1 | XP_016881772.1 | 1 | 311 | |||
GenBank mRNA | AK131444.1 | - | 1 | 676 | ||
BC031227.1 | - | 1 | 317 |