Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ?
Create Account
Products
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Tubes
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO 和 CRO 服務
CDMO 服務
臨床試驗 CRO 服務
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
Register for an Account
How to Order
Instrument Support
Technical Support Centers
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
快速訂購
登入
登入
Don't have an account ?
Create Account
帳戶
訂單
訂製產品與專案
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MEIS2 SIRNA Assays ›
Gene Symbol:
MEIS2
Gene Name:
Meis homeobox 2
Gene Aliases:
HsT18361, MRG1
Chromosome Location:
Chr.15: 36891021 - 37101299 on Build GRCh38
UniGene ID:
Hs.510989
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
40645
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
新增到購物車
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MEIS2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001220482.1
NP_001207411.1
6, 7
1038
NM_002399.3
NP_002390.1
5, 6
893
NM_170674.4
NP_733774.1
5, 6
1171
NM_170675.4
NP_733775.1
5, 6
1171
NM_170676.4
NP_733776.1
5, 6
1171
NM_170677.4
NP_733777.1
5, 6
1171
NM_172315.2
NP_758526.1
4, 5
647
NM_172316.2
NP_758527.1
5, 6
958
NR_051953.1
5, 6
1171
XM_006720523.1
XP_006720586.1
5, 6
561
XM_006720524.1
XP_006720587.1
5, 6
561
XM_006720525.1
XP_006720588.1
5, 6
561
XM_006720526.2
XP_006720589.1
5, 6
1557
XM_006720527.2
XP_006720590.1
4, 5
1438
XM_006720529.2
XP_006720592.1
4, 5
1438
XM_011521591.2
XP_011519893.1
3, 4
623
XM_017022205.1
XP_016877694.1
5, 6
1557
GenBank mRNA
AF178948.1
-
5, 6
752
AF179896.1
-
5, 6
752
AF179897.1
-
5, 6
752
AF179898.1
-
5, 6
752
AF179899.1
-
5, 6
752
AK055936.1
-
4, 5
371
AK056038.1
-
4, 5
588
AK056216.1
-
5, 6
1570
AK056620.1
-
5, 6
946
AK226072.1
-
5, 6
1174
AK294411.1
-
3, 4
325
AK300247.1
-
5, 6
926
AK311546.1
-
4, 5
467
BC001516.2
-
5, 6
897
2964406
BC001844.2
-
5, 6
897
2964406
BC007202.1
-
5, 6
897
2964406
BC050431.1
-
6, 7
918
6066728
Back To Top
More Information
Panther Classification:
Molecular Function -
homeodomain transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
eye development
transcription from RNA polymerase II promoter
visual learning
response to mechanical stimulus
pancreas development
negative regulation of myeloid cell differentiation
positive regulation of transcription from RNA polymerase II promoter
response to growth factor
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, sequence-specific DNA binding
transcription cofactor activity
transcription corepressor activity
protein binding
transcription factor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0