Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OVOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017020287.1 | XP_016875776.1 | 28 | 3767 | ||
GenBank mRNA | BX647938.1 | - | 9 | 6215 |
Gene Symbol: LOC728715 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017020286.1 | XP_016875775.1 | 3 | 317 |
Gene Symbol: OVOS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017020288.1 | XP_016875777.1 | 29 | 3947 | ||
GenBank mRNA | AL831947.1 | - | 1998 | |||
BC039117.1 | - | 2 | 372 | |||
BC046190.1 | - | 2 | 222 | |||
CR749395.1 | - | 2 | 2186 |