Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWA3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173615.4 | NP_775886.3 | 6, 7 | 591 | ||
GenBank mRNA | AK098260.1 | - | 5, 6 | 684 | ||
AK307544.1 | - | 6, 7 | 587 |