Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ENDOV SIRNA Assays ›
          Gene Symbol:
          ENDOV
          Gene Name:
          endonuclease V
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 80415117 - 80438086 on Build GRCh38
          UniGene ID:
          Hs.389678
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 41190

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ENDOV
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001164637.2 NP_001158109.1 4, 5 411
          NM_001164638.1 NP_001158110.1 4, 5 411
          NM_173627.4 NP_775898.2 5, 6 546
          XM_005257244.3 XP_005257301.1 5, 6 596
          XM_005257246.3 XP_005257303.1 4, 5 413
          XM_006721837.3 XP_006721900.1 5, 6 596
          XM_011524655.1 XP_011522957.1 5, 6 596
          XM_011524656.1 XP_011522958.1 5, 6 596
          XM_011524657.1 XP_011522959.1 5, 6 596
          XM_011524658.2 XP_011522960.1 5, 6 596
          XM_011524660.1 XP_011522962.1 4, 5 411
          XM_011524661.2 XP_011522963.1 4, 5 413
          XM_011524662.1 XP_011522964.1 5, 6 596
          XM_011524663.1 XP_011522965.1 5, 6 596
          XM_011524664.2 XP_011522966.1 6, 7 1214
          XM_011524666.1 XP_011522968.1 5, 6 778
          XM_011524667.2 XP_011522969.1 5, 6 724
          XM_011524669.2 XP_011522971.1 5, 6 596
          XM_011524670.1 XP_011522972.1 5, 6 596
          XM_011524671.2 XP_011522973.1 5, 6 596
          XM_011524672.1 XP_011522974.1 4, 5 394
          XM_011524673.1 XP_011522975.1 3, 4 351
          XM_011524674.1 XP_011522976.1 4, 5 394
          XM_011524675.1 XP_011522977.1 3, 4 371
          XM_011524676.1 XP_011522978.1 4, 5 643
          XM_011524677.1 XP_011522979.1 3, 4 354
          XM_011524678.2 XP_011522980.1 3, 4 509
          XM_017024508.1 XP_016879997.1 5, 6 1543
          XM_017024509.1 XP_016879998.1 5, 6 1489
          XM_017024510.1 XP_016879999.1 5, 6 596
          XM_017024511.1 XP_016880000.1 4, 5 413
          XM_017024512.1 XP_016880001.1 5, 6 724
          XM_017024513.1 XP_016880002.1 2, 3 469
          XM_017024514.1 XP_016880003.1 4, 5 394
          XM_017024515.1 XP_016880004.1 3, 4 354
          XM_017024516.1 XP_016880005.1 2, 3 257
          XM_017024517.1 XP_016880006.1 3, 4 371
          XM_017024518.1 XP_016880007.1 4, 5 394
          XM_017024519.1 XP_016880008.1 3, 4 354
          XM_017024520.1 XP_016880009.1 2, 3 680
          GenBank mRNA AK092539.1 - 4, 5 411
          AK096344.1 - 5, 6 546
          AK096802.1 - 3, 4 279
          AK123689.1 - 3, 4 354
          BC037889.2 - 2, 3 268
          BC045824.1 - 2, 3 268
          BC064545.1 - 4, 5 394 5736957
          BX647411.1 - 2, 3 274

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          DNA repair
          RNA phosphodiester bond hydrolysis, endonucleolytic
          magnesium ion binding
          single-stranded RNA binding
          endodeoxyribonuclease activity, producing 5'-phosphomonoesters
          endoribonuclease activity, producing 5'-phosphomonoesters
          structure-specific DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0