Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ST3GAL3 SIRNA Assays ›
Gene Symbol:
ST3GAL3
Gene Name:
ST3 beta-galactoside alpha-2,3-sialyltransferase 3
Gene Aliases:
EIEE15, MRT12, SIAT6, ST3GALII, ST3GalIII, ST3N
Chromosome Location:
Chr.1: 43707497 - 43931165 on Build GRCh38
UniGene ID:
Hs.597915
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
41404
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ST3GAL3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001270459.1
NP_001257388.1
12
1586
NM_001270460.1
NP_001257389.1
11
1583
NM_001270461.1
NP_001257390.1
9
1334
NM_001270462.1
NP_001257391.1
8
1241
NM_001270463.1
NP_001257392.1
8
1192
NM_001270464.1
NP_001257393.1
8
1147
NM_001270465.1
NP_001257394.1
8
1218
NM_001270466.1
NP_001257395.1
5
892
NM_006279.3
NP_006270.1
12
1676
NM_174963.3
NP_777623.2
13
1883
NM_174964.2
NP_777624.1
12
1721
NM_174965.2
NP_777625.1
7
1080
NM_174966.2
NP_777626.1
10
1382
NM_174967.2
NP_777627.1
7
1035
NM_174968.3
NP_777628.2
13
1838
NM_174969.2
NP_777629.1
11
1628
NM_174970.2
NP_777630.1
6
987
NM_174971.3
NP_777631.2
12
1790
NR_073016.1
12
1708
NR_073017.1
10
1585
NR_073018.1
11
1555
NR_073019.1
9
1524
NR_073020.1
10
1441
NR_073021.1
10
1369
NR_073023.1
9
1329
XM_006710825.3
XP_006710888.1
13
1908
XM_006710827.3
XP_006710890.1
12
1826
XM_006710828.3
XP_006710891.1
14
2048
XM_011541974.2
XP_011540276.1
14
2036
XM_011541976.2
XP_011540278.1
13
1988
XM_011541977.2
XP_011540279.1
13
1919
XM_011541980.2
XP_011540282.1
10
2139
XM_011541981.2
XP_011540283.1
11
1591
XM_011541983.2
XP_011540285.1
14
2183
XM_011541987.2
XP_011540289.1
14
2047
XM_011541988.2
XP_011540290.1
9
2064
XM_017002109.1
XP_016857598.1
14
2144
XM_017002111.1
XP_016857600.1
14
2137
XM_017002112.1
XP_016857601.1
14
2133
XM_017002113.1
XP_016857602.1
12
1756
XM_017002114.1
XP_016857603.1
14
2149
XM_017002115.1
XP_016857604.1
13
2000
XM_017002116.1
XP_016857605.1
13
1971
XM_017002117.1
XP_016857606.1
11
1546
XM_017002118.1
XP_016857607.1
10
1429
XM_017002119.1
XP_016857608.1
13
1850
XM_017002120.1
XP_016857609.1
13
1848
XM_017002121.1
XP_016857610.1
11
1641
XM_017002122.1
XP_016857611.1
13
2007
GenBank mRNA
AY051143.1
-
11
1613
BC050380.1
-
12
1660
5769575
Back To Top
More Information
Panther Classification:
Molecular Function -
transferase
metabolite interconversion enzyme
Gene Ontology Categories:
Function(s)
Process(es)
ganglioside biosynthetic process
oligosaccharide metabolic process
O-glycan processing
keratan sulfate biosynthetic process
protein N-linked glycosylation via asparagine
sialylation
beta-galactoside (CMP) alpha-2,3-sialyltransferase activity
N-acetyllactosaminide alpha-2,3-sialyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-cdcg2:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0