Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WFIKKN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175575.5 | NP_783165.1 | 1, 2 | 912 | ||
GenBank mRNA | AF468657.1 | - | 1, 2 | 234 | ||
AK127743.1 | - | 1, 2 | 572 | |||
AK312821.1 | - | 1, 2 | 406 | |||
AL537450.3 | - | 1, 2 | 675 | |||
AY358142.1 | - | 1, 2 | 747 | |||
BC140842.1 | - | 1, 2 | 306 | 9021710 | ||
BC140844.1 | - | 1, 2 | 306 | 9021712 |