Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LPAR1 SIRNA Assays ›
Gene Symbol:
LPAR1
Gene Name:
lysophosphatidic acid receptor 1
Gene Aliases:
EDG2, GPR26, Gpcr26, LPA1, Mrec1.3, VZG1, edg-2, rec.1.3, vzg-1
Chromosome Location:
Chr.9: 110873252 - 111039259 on Build GRCh38
UniGene ID:
Hs.126667
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
4144
Validated
Purification
HPLC
Standard
In-Vivo Ready
Size
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
Availability
Inventoried
Catalog #
AM51331
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LPAR1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001401.3
NP_001392.2
4
1829
NM_057159.2
NP_476500.1
5
1907
XM_005251781.3
XP_005251838.1
5
2052
XM_005251782.3
XP_005251839.1
5
2146
XM_017014383.1
XP_016869872.1
5
1946
XM_017014384.1
XP_016869873.1
6
2320
XM_017014385.1
XP_016869874.1
7
1913
XM_017014386.1
XP_016869875.1
7
2213
XM_017014387.1
XP_016869876.1
7
1949
XM_017014388.1
XP_016869877.1
7
2132
XM_017014389.1
XP_016869878.1
6
1875
XM_017014390.1
XP_016869879.1
7
1910
XM_017014391.1
XP_016869880.1
7
1934
XM_017014392.1
XP_016869881.1
6
2177
XM_017014393.1
XP_016869882.1
7
1973
XM_017014394.1
XP_016869883.1
6
2102
XM_017014395.1
XP_016869884.1
6
1930
XM_017014396.1
XP_016869885.1
5
1872
XM_017014397.1
XP_016869886.1
6
1803
XM_017014398.1
XP_016869887.1
6
2046
XM_017014399.1
XP_016869888.1
6
1801
XM_017014400.1
XP_016869889.1
7
2162
XM_017014401.1
XP_016869890.1
6
1833
XM_017014402.1
XP_016869891.1
6
1890
XM_017014403.1
XP_016869892.1
7
1888
XM_017014404.1
XP_016869893.1
5
2086
XM_017014405.1
XP_016869894.1
7
2075
XM_017014406.1
XP_016869895.1
6
1809
XM_017014407.1
XP_016869896.1
6
1865
GenBank mRNA
AK022808.1
-
4
1829
BC030615.2
-
5
1736
4815446
BC036034.1
-
5
1866
5272431
BC073167.1
-
6
1776
6193969
U80811.1
-
3
1507
Back To Top
More Information
Set Membership:
G-protein coupled receptor
Matching Taqman Gene Expression Assay:
Hs00173500_m1
Panther Classification:
Molecular Function -
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPK activity
G-protein coupled receptor signaling pathway
adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
activation of phospholipase C activity
positive regulation of cytosolic calcium ion concentration
regulation of cell shape
negative regulation of neuron projection development
oligodendrocyte development
cerebellum development
optic nerve development
corpus callosum development
negative regulation of cAMP biosynthetic process
bleb assembly
positive regulation of Rho protein signal transduction
myelination
positive regulation of apoptotic process
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of MAPK cascade
positive regulation of cytosolic calcium ion concentration involved in phospholipase C-activating G-protein coupled signaling pathway
positive regulation of stress fiber assembly
cell chemotaxis
positive regulation of dendritic spine development
cellular response to oxygen levels
positive regulation of smooth muscle cell chemotaxis
cellular response to 1-oleoyl-sn-glycerol 3-phosphate
G-protein alpha-subunit binding
G-protein coupled receptor activity
protein binding
PDZ domain binding
lysophosphatidic acid binding
lysophosphatidic acid receptor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-89mxr:80/10.239.163.131:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0