Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPIFA4P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026760.1 | 4 | 368 | |||
GenBank mRNA | AY180924.1 | - | 4 | 368 | ||
AY389991.1 | - | 3 | 337 | |||
AY389992.1 | - | 3 | 280 | |||
BC069128.1 | - | 3 | 331 | |||
BC069315.1 | - | 4 | 367 |