Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287529.1 | NP_001274458.1 | 10 | 1874 | ||
NM_174983.4 | NP_778148.2 | 10 | 1901 | |||
XM_017026288.1 | XP_016881777.1 | 9 | 1737 | |||
GenBank mRNA | BC036706.2 | - | 10 | 1853 | 4623094 | |
BC068439.1 | - | 10 | 1890 | 5314853 | ||
BC094804.1 | - | 2 | 331 |