Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FKBP1A SIRNA Assays ›
          Gene Symbol:
          FKBP1A FKBP1AP1
          Gene Name:
          FK506 binding protein 1A
          FK506 binding protein 1A pseudogene 1
          Gene Aliases:
          FKBP-12, FKBP-1A, FKBP1, FKBP12, PKC12, PKCI2, PPIASE
          FKBP12, FKBP1P1
          Chromosome Location:
          Chr.20: 1368977 - 1393172 on Build GRCh38
          Chromosome Location:
          Chr.19: 57825280 - 57827462 on Build GRCh38
          UniGene ID:
          Hs.471933
          Hs.720356
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 41616

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FKBP1A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000801.4 NP_000792.1 3 287
          NM_054014.3 NP_463460.1 3 287
          GenBank mRNA AB241123.1 - 2 65
          AB241125.1 - 4 195
          BC001925.1 - 3 161 2823635
          BC005147.2 - 3 144 3504715
          BC119732.1 - 3 224 40118164
          BT007066.1 - 3 113
          CR407613.1 - 3 113
          CR542168.1 - 3 113
          CX870892.1 - 3 287
          M34539.1 - 3 191
          X52220.1 - 3 143
          Gene Symbol: FKBP1AP1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_024162.1 2 276
          GenBank mRNA AI076645.1 - 2 276

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          protein peptidyl-prolyl isomerization
          heart morphogenesis
          protein folding
          'de novo' protein folding
          transforming growth factor beta receptor signaling pathway
          SMAD protein complex assembly
          protein maturation by protein folding
          positive regulation of protein ubiquitination
          positive regulation of protein binding
          negative regulation of protein phosphatase type 2B activity
          regulation of protein localization
          regulation of activin receptor signaling pathway
          protein refolding
          T cell activation
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          extracellular fibril organization
          regulation of immune response
          negative regulation of release of sequestered calcium ion into cytosol
          ventricular cardiac muscle tissue morphogenesis
          regulation of ryanodine-sensitive calcium-release channel activity
          negative regulation of ryanodine-sensitive calcium-release channel activity
          heart trabecula formation
          chaperone-mediated protein folding
          calcium ion transmembrane transport
          regulation of amyloid precursor protein catabolic process
          amyloid fibril formation
          peptidyl-prolyl cis-trans isomerase activity
          signal transducer activity
          transforming growth factor beta receptor binding
          protein binding
          macrolide binding
          FK506 binding
          calcium channel inhibitor activity
          type I transforming growth factor beta receptor binding
          ion channel binding
          SMAD binding
          activin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0