Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ASCL1 SIRNA Assays ›
          Gene Symbol:
          ASCL1
          Gene Name:
          achaete-scute family bHLH transcription factor 1
          Gene Aliases:
          ASH1, HASH1, MASH1, bHLHa46
          Chromosome Location:
          Chr.12: 102957674 - 102960516 on Build GRCh38
          UniGene ID:
          Hs.703025
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 42069

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ASCL1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_004316.3 NP_004307.2 1 1038
          GenBank mRNA AK290539.1 - 1 1039
          AK314114.1 - 1 654
          BC001638.2 - 1 1026
          BC002341.2 - 1 1020 2821173
          BC003134.1 - 1 1020 2821173
          BC004425.1 - 1 1026 2819879
          BC031299.1 - 1 1004 3534115
          L08424.1 - 1 905

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          basic helix-loop-helix transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          neuron migration
          noradrenergic neuron development
          noradrenergic neuron fate commitment
          transcription, DNA-templated
          Notch signaling pathway
          regulation of mitotic cell cycle
          neuroblast fate determination
          neuroblast proliferation
          response to lithium ion
          regulation of gene expression
          oligodendrocyte development
          spinal cord association neuron differentiation
          spinal cord oligodendrocyte cell fate specification
          vestibular nucleus development
          cerebral cortex GABAergic interneuron differentiation
          commitment of neuronal cell to specific neuron type in forebrain
          central nervous system neuron development
          cerebral cortex development
          neurogenesis
          regulation of epithelial cell differentiation
          response to retinoic acid
          negative regulation of apoptotic process
          positive regulation of neuron apoptotic process
          negative regulation of neuron differentiation
          positive regulation of neuron differentiation
          positive regulation of Notch signaling pathway
          positive regulation of cell cycle
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          cell maturation
          sympathetic nervous system development
          neuron fate specification
          musculoskeletal movement, spinal reflex action
          response to folic acid
          subpallium neuron fate commitment
          regulation of timing of subpallium neuron differentiation
          olfactory pit development
          lung epithelial cell differentiation
          ventral spinal cord interneuron fate commitment
          lung neuroendocrine cell differentiation
          stomach neuroendocrine cell differentiation
          carotid body glomus cell differentiation
          adrenal chromaffin cell differentiation
          sympathetic ganglion development
          response to epidermal growth factor
          cellular response to magnetism
          positive regulation of neural precursor cell proliferation
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcription factor activity, transcription factor binding
          transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          protein homodimerization activity
          bHLH transcription factor binding
          E-box binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0