Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HLA-B SIRNA Assays ›
          Gene Symbol:
          HLA-B
          Gene Name:
          major histocompatibility complex, class I, B
          Gene Aliases:
          AS, B-4901, B-5001, Bw-47, Bw-50, HEL-S-83, HLA-B*45ZJ, HLA-B-3506, HLA-B-3905, HLA-B-5502, HLA-B-5602, HLA-B15, HLA-B39, HLA-B49, HLA-B50, HLA-B55, HLA-B59, HLA-B61, HLA-Cw, HLA-DRB1, HLAB, SPDA1
          Chromosome Location:
          Chr.6: 31353866 - 31357245 on Build GRCh38
          UniGene ID:
          Hs.654404
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 42112

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HLA-B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_005514.7 NP_005505.2 4 868
          XM_011514557.1 XP_011512859.1 4 791
          GenBank mRNA AB008102.1 - 4 781
          AF006634.1 - 4 781
          AF016641.1 - 4 781
          AF022783.1 - 4 781
          AF026218.1 - 4 781
          AF130734.1 - 5 781
          AF170577.1 - 4 781
          AF189017.1 - 4 781
          AF389378.1 - 4 785
          AJ131118.1 - 4 781
          AJ251593.1 - 4 780
          AJ506054.1 - 1 162
          AJ535113.1 - 4 785
          AJ634528.1 - 4 781
          AK026595.1 - 5 804
          AK124160.1 - 4 1051
          AK129796.1 - 4 802
          AK130137.1 - 4 802
          AK130155.1 - 4 804
          AK130166.1 - 4 802
          AK130169.1 - 4 808
          AK130654.1 - 5 741
          AK290570.1 - 4 802
          AK292226.1 - 4 802
          AK300685.1 - 5 711
          AK313911.1 - 4 802
          AM711637.1 - 4 781
          AY007140.1 - 4 801
          AY221104.1 - 3 685
          AY569160.1 - 3 708
          BC007243.1 - 5 775
          BC013187.1 - 4 790 4344323
          BC091497.1 - 4 785 30386550
          BX647315.1 - 5 803
          D14343.1 - 4 745
          D25275.1 - 4 781
          D31816.1 - 4 781
          D49824.1 - 4 781
          D50290.1 - 4 781
          D50291.1 - 4 781
          D50292.1 - 4 781
          D50293.1 - 4 781
          D50294.1 - 4 781
          D50295.1 - 4 781
          D50296.1 - 4 781
          D50299.1 - 4 781
          D50300.1 - 4 781
          D83043.1 - 4 781
          D85761.1 - 5 781
          D85762.1 - 4 781
          D87665.1 - 5 772
          DQ336696.1 - 4 769
          DQ336697.1 - 4 775
          DQ336698.1 - 4 769
          EU048339.1 - 1 162
          EU668336.1 - 4 781
          FM205711.1 - 4 781
          HE663398.1 - 5 730
          HG327115.1 - 4 781
          HG327117.1 - 4 781
          HG327119.1 - 4 781
          HG327120.1 - 4 781
          HG514457.1 - 4 781
          KU204955.1 - 2 438
          L04695.1 - 4 781
          L04696.1 - 4 781
          L07743.1 - 4 781
          L07950.1 - 4 781
          L09735.1 - 4 781
          L09736.1 - 4 781
          L11570.1 - 4 781
          L11571.1 - 4 781
          L11603.1 - 4 781
          L11604.1 - 4 781
          L11666.1 - 4 781
          L15005.1 - 4 781
          L19923.1 - 4 781
          L19937.1 - 4 781
          L20089.1 - 4 776
          L20090.1 - 5 776
          L22027.1 - 4 781
          L32862.1 - 4 781
          L33922.1 - 4 781
          L37880.1 - 4 781
          L37881.1 - 4 781
          L38504.1 - 4 781
          L41214.1 - 5 781
          L41925.1 - 5 781
          L42345.1 - 4 781
          M12678.1 - 4 772
          M15470.1 - 4 772
          M16102.1 - 4 788
          M24033.1 - 4 781
          M27540.1 - 3 595
          M28203.1 - 4 770
          M28204.1 - 4 745
          M28205.1 - 5 781
          M32317.1 - 4 781
          M32318.1 - 5 781
          M32319.1 - 5 781
          M32320.1 - 4 781
          M62852.1 - 3 709
          M75138.1 - 4 781
          M77774.1 - 5 781
          M77775.1 - 4 781
          M77776.1 - 4 781
          M77777.1 - 4 781
          M77778.1 - 4 781
          M81798.1 - 4 781
          M83191.1 - 3 709
          M83192.1 - 3 709
          M83193.1 - 3 709
          M83194.1 - 3 709
          M83195.1 - 3 709
          M84380.1 - 4 781
          M84381.1 - 4 781
          M84382.1 - 4 781
          M84383.1 - 4 781
          M84384.1 - 4 781
          M84385.1 - 4 781
          M84694.1 - 4 781
          M86403.1 - 4 757
          U01848.1 - 4 781
          U03027.1 - 4 781
          U03698.1 - 4 781
          U03859.1 - 4 781
          U04244.1 - 4 781
          U04245.1 - 4 781
          U06697.1 - 3 708
          U06862.1 - 4 781
          U09864.1 - 4 781
          U09912.1 - 4 781
          U11261.1 - 4 781
          U11262.1 - 4 781
          U11263.1 - 4 781
          U11264.1 - 4 781
          U11265.1 - 4 781
          U11266.1 - 4 781
          U11267.1 - 4 781
          U11268.1 - 4 781
          U11269.1 - 4 781
          U14943.1 - 4 781
          U17107.1 - 4 781
          U17572.1 - 4 781
          U21052.1 - 4 781
          U21053.1 - 4 781
          U29057.1 - 4 781
          U30904.1 - 4 781
          U30936.1 - 4 781
          U31971.1 - 4 781
          U35431.1 - 4 781
          U35734.1 - 4 781
          U36392.1 - 4 781
          U36492.1 - 4 781
          U38800.1 - 4 781
          U50710.1 - 4 781
          U57966.1 - 4 781
          U63653.1 - 4 781
          U64801.1 - 4 781
          U70528.1 - 4 781
          U70529.1 - 4 781
          U75533.1 - 4 781
          U80945.1 - 4 781
          U88407.1 - 4 781
          V00527.1 - 1 44
          X61706.1 - 4 781
          X61708.1 - 5 781
          X61709.1 - 4 781
          X61710.1 - 4 781
          X64366.1 - 4 781
          X64454.1 - 4 781
          X73578.1 - 4 781
          X75953.1 - 5 781
          X81363.1 - 4 781
          X84725.1 - 4 781
          X90390.1 - 4 781
          X90391.1 - 4 781
          X91749.1 - 4 781
          Y08994.1 - 5 781
          Y08995.1 - 4 781
          Y09118.1 - 4 781
          Y13567.1 - 4 781
          Z15143.1 - 5 781
          Z22651.1 - 4 781
          Z33453.1 - 4 782

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          major histocompatibility complex protein

          Gene Ontology Categories:

          Function(s) Process(es)

          antigen processing and presentation of peptide antigen via MHC class I
          antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-dependent
          antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-independent
          viral process
          antigen processing and presentation
          regulation of immune response
          interferon-gamma-mediated signaling pathway
          type I interferon signaling pathway
          antigen binding
          peptide antigen binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0