Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TUBA1B SIRNA Assays ›
          Gene Symbol:
          TUBA1B
          Gene Name:
          tubulin alpha 1b
          Gene Aliases:
          K-ALPHA-1
          Chromosome Location:
          Chr.12: 49127782 - 49131521 on Build GRCh38
          UniGene ID:
          Hs.524390
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 42217

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TUBA1B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_006082.2 NP_006073.2 4 1574
          GenBank mRNA AK026516.1 - 4 1448
          AK026632.1 - 4 1448
          AK056693.1 - 3 2356
          AK057773.1 - 4 1448
          AK094717.1 - 3 3021
          AK098357.1 - 4 1448
          AK098635.1 - 4 1448
          AK098771.1 - 4 1448
          AK289354.1 - 4 1448
          AK293337.1 - 2 1073
          BC000696.2 - 4 1439 3349131
          BC001128.1 - 4 1432 2964447
          BC006379.2 - 4 1434 4100566
          BC006481.1 - 4 1434 2905305
          BC008659.2 - 4 1411 3871729
          BC009314.2 - 4 1433 4111591
          BC009509.2 - 4 1435 3533174
          BC009512.2 - 4 1432 3537301
          BC009513.2 - 4 1436 3537827
          BC010494.1 - 4 1434 3445903
          BC011572.1 - 4 1434 4561240
          BC015883.2 - 4 1392 3909374
          BC017004.1 - 4 1404 4430457
          BC018948.2 - 4 1434
          BC021564.1 - 4 1086 4579604
          BC030820.1 - 4 1445 4277263
          BC071904.1 - 4 1439 6089096
          DQ400107.1 - 4 1438
          K00558.1 - 4 1420
          Z36832.1 - 1 660

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          tubulin

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule cytoskeleton organization
          microtubule-based process
          cytoskeleton-dependent intracellular transport
          cell division
          cellular response to interleukin-4
          double-stranded RNA binding
          GTPase activity
          structural molecule activity
          structural constituent of cytoskeleton
          protein binding
          GTP binding
          ubiquitin protein ligase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline