siRNA Details
Gene Symbol: SOX21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007084.3 | NP_009015.1 | 1 | 766 | ||
GenBank mRNA | DB493621.1 | - | 1 | 317 |

Gene Symbol: SOX21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007084.3 | NP_009015.1 | 1 | 766 | ||
GenBank mRNA | DB493621.1 | - | 1 | 317 |