Search
Search

RTN4R| Gene Symbol: RTN4R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023004.5 | NP_075380.1 | 2 | 536 | ||
| GenBank mRNA | AF283463.1 | - | 2 | 347 | ||
| AK054602.1 | - | 2 | 592 | |||
| AL834449.1 | - | 2 | 479 | |||
| BC011787.1 | - | 2 | 381 | 4040540 | ||
| CR456360.1 | - | 2 | 375 | |||