Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FIBCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145106.1 | NP_001138578.1 | 8 | 1664 | ||
NM_032843.4 | NP_116232.3 | 7 | 1802 | |||
GenBank mRNA | AK027716.1 | - | 5 | 987 | ||
AK292489.1 | - | 7 | 1571 | |||
AY358311.1 | - | 7 | 1559 | |||
BC032953.2 | - | 7 | 1800 | 4811679 | ||
BC112892.1 | - | 7 | 1552 |