Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PTPN2 SIRNA Assays ›
          Gene Symbol:
          PTPN2
          Gene Name:
          protein tyrosine phosphatase, non-receptor type 2
          Gene Aliases:
          PTN2, PTPT, TC-PTP, TCELLPTP, TCPTP
          Chromosome Location:
          Chr.18: 12785478 - 12884351 on Build GRCh38
          UniGene ID:
          Hs.654527
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 4265

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PTPN2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001207013.1 NP_001193942.1 3 436
          NM_001308287.1 NP_001295216.1 2 331
          NM_002828.3 NP_002819.2 3 436
          NM_080422.2 NP_536347.1 3 436
          NM_080423.2 NP_536348.1 3 436
          XM_005258124.3 XP_005258181.1 3 434
          XM_005258125.3 XP_005258182.1 3 434
          XM_011525705.2 XP_011524007.1 4 654
          XM_011525706.2 XP_011524008.1 3 359
          XM_017025884.1 XP_016881373.1 3 452
          XM_017025885.1 XP_016881374.1 4 638
          XM_017025886.1 XP_016881375.1 2 326
          XM_017025887.1 XP_016881376.1 2 277
          XM_017025888.1 XP_016881377.1 2 277
          GenBank mRNA AB209569.1 - 2 363
          AK225706.1 - 3 315
          AK290649.1 - 3 318
          AK292570.1 - 3 336
          BC008244.1 - 3 279 3872164
          BC016727.1 - 3 348 4081330
          BG699440.1 - 3 441
          M25393.1 - 3 302

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          protein phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of cell proliferation
          insulin receptor signaling pathway
          negative regulation of tumor necrosis factor-mediated signaling pathway
          negative regulation of lipid storage
          B cell differentiation
          T cell differentiation
          erythrocyte differentiation
          peptidyl-tyrosine dephosphorylation
          negative regulation of epidermal growth factor receptor signaling pathway
          negative regulation of tyrosine phosphorylation of Stat1 protein
          negative regulation of tyrosine phosphorylation of Stat5 protein
          negative regulation of tyrosine phosphorylation of Stat6 protein
          glucose homeostasis
          negative regulation of macrophage differentiation
          positive regulation of gluconeogenesis
          negative regulation of insulin receptor signaling pathway
          negative regulation of inflammatory response
          negative regulation of T cell receptor signaling pathway
          negative regulation of chemotaxis
          regulation of interferon-gamma-mediated signaling pathway
          negative regulation of interferon-gamma-mediated signaling pathway
          negative regulation of type I interferon-mediated signaling pathway
          negative regulation of protein tyrosine kinase activity
          negative regulation of interleukin-6-mediated signaling pathway
          negative regulation of ERK1 and ERK2 cascade
          regulation of hepatocyte growth factor receptor signaling pathway
          negative regulation of interleukin-2-mediated signaling pathway
          negative regulation of prolactin signaling pathway
          negative regulation of interleukin-4-mediated signaling pathway
          negative regulation of macrophage colony-stimulating factor signaling pathway
          negative regulation of positive thymic T cell selection
          positive regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
          positive regulation of PERK-mediated unfolded protein response
          negative regulation of platelet-derived growth factor receptor-beta signaling pathway
          protein tyrosine phosphatase activity
          non-membrane spanning protein tyrosine phosphatase activity
          integrin binding
          protein binding
          protein kinase binding
          syntaxin binding
          receptor tyrosine kinase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-49ts5:80/100.66.72.225:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0