Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_058173.2 | NP_477521.1 | 3, 4 | 290 | ||
GenBank mRNA | AF414087.1 | - | 3, 4 | 268 | ||
AY359062.1 | - | 3, 4 | 261 | |||
AY651259.1 | - | 4, 5 | 198 | |||
BC111421.1 | - | 3, 4 | 255 | 40021582 |