Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PTPRD SIRNA Assays ›
Gene Symbol:
PTPRD
Gene Name:
protein tyrosine phosphatase, receptor type D
Gene Aliases:
HPTP, HPTPD, HPTPDELTA, PTPD, RPTPDELTA
Chromosome Location:
Chr.9: 8314246 - 10612840 on Build GRCh38
UniGene ID:
Hs.446083
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
4273
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PTPRD
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001040712.2
NP_001035802.1
2
185
NM_001171025.1
NP_001164496.1
2
185
NM_002839.3
NP_002830.1
13
825
NM_130391.3
NP_569075.2
2
185
NM_130392.3
NP_569076.2
2
185
NM_130393.3
NP_569077.2
2
185
XM_006716817.3
XP_006716880.1
12
1123
XM_006716823.2
XP_006716886.1
12
1117
XM_006716825.3
XP_006716888.1
12
1119
XM_006716827.3
XP_006716890.1
12
1116
XM_006716832.3
XP_006716895.1
12
1111
XM_006716833.3
XP_006716896.1
12
1113
XM_006716834.3
XP_006716897.1
12
1114
XM_006716835.3
XP_006716898.1
12
1113
XM_006716837.3
XP_006716900.1
12
1111
XM_006716838.3
XP_006716901.1
12
1113
XM_006716839.3
XP_006716902.1
12
1111
XM_011517992.2
XP_011516294.1
12
1116
XM_017014958.1
XP_016870447.1
12
1122
XM_017014959.1
XP_016870448.1
13
1194
XM_017014960.1
XP_016870449.1
11
1083
XM_017014961.1
XP_016870450.1
11
1067
XM_017014962.1
XP_016870451.1
3
255
XM_017014963.1
XP_016870452.1
12
1122
XM_017014964.1
XP_016870453.1
12
1121
XM_017014965.1
XP_016870454.1
12
1124
XM_017014966.1
XP_016870455.1
12
1122
XM_017014967.1
XP_016870456.1
12
1120
XM_017014968.1
XP_016870457.1
12
1119
XM_017014969.1
XP_016870458.1
12
1122
XM_017014970.1
XP_016870459.1
12
1122
XM_017014971.1
XP_016870460.1
12
1120
XM_017014972.1
XP_016870461.1
12
1122
XM_017014973.1
XP_016870462.1
12
1119
XM_017014974.1
XP_016870463.1
12
1122
XM_017014975.1
XP_016870464.1
12
1117
XM_017014976.1
XP_016870465.1
12
1119
XM_017014977.1
XP_016870466.1
12
1120
XM_017014978.1
XP_016870467.1
12
1119
XM_017014979.1
XP_016870468.1
12
1121
XM_017014980.1
XP_016870469.1
12
1116
XM_017014981.1
XP_016870470.1
12
1111
XM_017014982.1
XP_016870471.1
12
1118
XM_017014983.1
XP_016870472.1
12
1118
XM_017014984.1
XP_016870473.1
12
1118
XM_017014985.1
XP_016870474.1
12
1115
XM_017014986.1
XP_016870475.1
12
1114
XM_017014987.1
XP_016870476.1
12
1114
XM_017014988.1
XP_016870477.1
12
1114
XM_017014989.1
XP_016870478.1
12
1114
XM_017014990.1
XP_016870479.1
12
1113
XM_017014991.1
XP_016870480.1
12
1113
XM_017014992.1
XP_016870481.1
12
1113
XM_017014993.1
XP_016870482.1
12
1111
XM_017014994.1
XP_016870483.1
12
1111
XM_017014995.1
XP_016870484.1
12
1117
GenBank mRNA
AB208869.1
-
11
AB211400.1
-
12
793
BC028038.1
-
897
BC106713.1
-
2
133
40027581
BC106714.1
-
2
133
40027582
BC106715.2
-
2
133
40027584
BC106716.1
-
2
133
40027585
DR001889.1
-
4
260
L38929.1
-
3
235
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein dephosphorylation
phosphate-containing compound metabolic process
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
transmembrane receptor protein tyrosine phosphatase signaling pathway
chemical synaptic transmission
neuron differentiation
peptidyl-tyrosine dephosphorylation
positive regulation of dendrite morphogenesis
presynaptic membrane assembly
transmembrane receptor protein tyrosine phosphatase activity
receptor binding
protein binding
cell adhesion molecule binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7dbcb7bc98-zbtnl:80/10.239.166.103:80.
git-commit: 8789124447060be43cea05543d36199cab59d3d8
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.5.0-2024.03.10-1.0