Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YPEL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145524.1 | NP_001138996.1 | 5 | 506 | ||
NM_031477.4 | NP_113665.3 | 4 | 1152 | |||
GenBank mRNA | AB098737.1 | - | 5 | 506 | ||
AF305622.1 | - | 5 | 509 | |||
BC005009.1 | - | 5 | 462 | 3639517 | ||
BC050664.1 | - | 4 | 1151 | 5748830 |