Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PML SIRNA Assays ›
          Gene Symbol:
          PML
          Gene Name:
          promyelocytic leukemia
          Gene Aliases:
          MYL, PP8675, RNF71, TRIM19
          Chromosome Location:
          Chr.15: 73994673 - 74047819 on Build GRCh38
          UniGene ID:
          Hs.526464
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 42781

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PML
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002675.3 NP_002666.1 3, 4 1331
          NM_033238.2 NP_150241.2 3, 4 1331
          NM_033239.2 NP_150242.1 3, 4 1331
          NM_033240.2 NP_150243.2 3, 4 1331
          NM_033244.3 NP_150247.2 3, 4 1331
          NM_033246.2 NP_150249.1 3, 4 1331
          NM_033247.2 NP_150250.2 3, 4 1331
          NM_033249.2 NP_150252.1 3, 4 1331
          NM_033250.2 NP_150253.2 3, 4 1331
          GenBank mRNA AB208950.1 - 3, 4 1201
          AB209051.1 - 2, 3 1042
          AB209411.1 - 3, 4 1275
          AF230401.1 - 3, 4 1263
          AF230402.1 - 3, 4 1263
          AF230403.1 - 3, 4 1263
          AF230404.1 - 3, 4 1263
          AF230405.1 - 3, 4 1263
          AF230406.1 - 3, 4 1263
          AF230407.1 - 3, 4 1263
          AF230408.1 - 3, 4 1263
          AF230409.1 - 3, 4 1263
          AF230410.1 - 3, 4 1263
          AF230411.1 - 3, 4 1263
          BC000080.2 - 3, 4 1252 3506984
          BC020994.2 - 3, 4 1252 2958462
          BT009911.1 - 3, 4 1191
          EF535849.1 - 64
          M73778.1 - 3, 4 1271
          M79462.1 - 3, 4 1205
          M79463.1 - 3, 4 1205
          M79464.1 - 3, 4 1205
          M80185.1 - 3, 4 1205
          S50913.1 - 3, 4 1273
          X63131.1 - 3, 4 1332

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          regulation of protein phosphorylation
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          protein complex assembly
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
          cell cycle arrest
          transforming growth factor beta receptor signaling pathway
          common-partner SMAD protein phosphorylation
          SMAD protein import into nucleus
          negative regulation of cell proliferation
          intrinsic apoptotic signaling pathway in response to oxidative stress
          response to UV
          response to gamma radiation
          regulation of calcium ion transport into cytosol
          fibroblast migration
          negative regulation of angiogenesis
          protein sumoylation
          myeloid cell differentiation
          regulation of cell adhesion
          negative regulation of cell growth
          PML body organization
          positive regulation of telomere maintenance
          endoplasmic reticulum calcium ion homeostasis
          circadian regulation of gene expression
          response to cytokine
          regulation of circadian rhythm
          intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
          entrainment of circadian clock by photoperiod
          proteasome-mediated ubiquitin-dependent protein catabolic process
          innate immune response
          cell fate commitment
          regulation of MHC class I biosynthetic process
          negative regulation of transcription, DNA-templated
          positive regulation of fibroblast proliferation
          retinoic acid receptor signaling pathway
          negative regulation of interleukin-1 beta secretion
          maintenance of protein location in nucleus
          defense response to virus
          interferon-gamma-mediated signaling pathway
          branching involved in mammary gland duct morphogenesis
          intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
          cellular response to interleukin-4
          cellular senescence
          extrinsic apoptotic signaling pathway
          regulation of signal transduction by p53 class mediator
          negative regulation of viral release from host cell
          negative regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          regulation of double-strand break repair
          positive regulation of extrinsic apoptotic signaling pathway
          DNA binding
          protein binding
          zinc ion binding
          ubiquitin protein ligase binding
          SUMO binding
          protein homodimerization activity
          SMAD binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0