Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other EGFR SIRNA Assays ›
          Gene Symbol:
          EGFR
          Gene Name:
          epidermal growth factor receptor
          Gene Aliases:
          ERBB, ERBB1, HER1, NISBD2, PIG61, mENA
          Chromosome Location:
          Chr.7: 55019032 - 55207338 on Build GRCh38
          UniGene ID:
          Hs.488293
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 42833

          Purification
          Size
          Availability Inventoried
          Catalog # AM51331
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: EGFR
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_005228.3 NP_005219.2 3 653
          NM_201282.1 NP_958439.1 3 653
          NM_201283.1 NP_958440.1 3 653
          NM_201284.1 NP_958441.1 3 653
          GenBank mRNA AB209442.1 - 3 553
          AF125253.1 - 3 652
          AK127817.1 - 1 951
          AK225422.1 - 3 568
          AK290352.1 - 3 453
          AK294750.1 - 3 431
          AY550966.1 - 3 453
          AY698023.1 - 3 407
          AY698024.1 - 3 407
          BC094761.1 - 3 664 30528231
          DQ088980.1 - 3 453
          GU255993.1 - 3 450
          HQ912715.1 - 3 653
          K03193.1 - 3 566
          U48722.1 - 3 651
          U95089.1 - 4 910
          X00588.1 - 3 593

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          activation of MAPKK activity
          cell morphogenesis
          ossification
          embryonic placenta development
          positive regulation of protein phosphorylation
          hair follicle development
          response to stress
          signal transduction
          cell surface receptor signaling pathway
          epidermal growth factor receptor signaling pathway
          activation of phospholipase C activity
          salivary gland morphogenesis
          learning or memory
          cell proliferation
          positive regulation of cell proliferation
          regulation of phosphatidylinositol 3-kinase signaling
          single organismal cell-cell adhesion
          peptidyl-tyrosine phosphorylation
          cerebral cortex cell migration
          positive regulation of cell growth
          positive regulation of cell migration
          positive regulation of cyclin-dependent protein serine/threonine kinase activity involved in G1/S transition of mitotic cell cycle
          positive regulation of catenin import into nucleus
          ERBB2 signaling pathway
          negative regulation of epidermal growth factor receptor signaling pathway
          negative regulation of protein catabolic process
          positive regulation of phosphorylation
          activation of phospholipase A2 activity by calcium-mediated signaling
          negative regulation of apoptotic process
          positive regulation of MAP kinase activity
          positive regulation of GTPase activity
          positive regulation of nitric oxide biosynthetic process
          positive regulation of DNA repair
          positive regulation of DNA replication
          positive regulation of transcription from RNA polymerase II promoter
          protein autophosphorylation
          phosphatidylinositol phosphorylation
          phosphatidylinositol-mediated signaling
          positive regulation of fibroblast proliferation
          digestive tract morphogenesis
          positive regulation of epithelial cell proliferation
          regulation of peptidyl-tyrosine phosphorylation
          regulation of nitric-oxide synthase activity
          protein insertion into membrane
          positive regulation of protein kinase B signaling
          morphogenesis of an epithelial fold
          eyelid development in camera-type eye
          response to UV-A
          positive regulation of ERK1 and ERK2 cascade
          cellular response to amino acid stimulus
          cellular response to epidermal growth factor stimulus
          cellular response to estradiol stimulus
          positive regulation of production of miRNAs involved in gene silencing by miRNA
          regulation of cell motility
          chromatin binding
          double-stranded DNA binding
          MAP kinase kinase kinase activity
          protein tyrosine kinase activity
          transmembrane receptor protein tyrosine kinase activity
          receptor signaling protein tyrosine kinase activity
          transmembrane signaling receptor activity
          epidermal growth factor-activated receptor activity
          Ras guanyl-nucleotide exchange factor activity
          protein binding
          ATP binding
          enzyme binding
          protein phosphatase binding
          nitric-oxide synthase regulator activity
          ubiquitin protein ligase binding
          identical protein binding
          phosphatidylinositol-4,5-bisphosphate 3-kinase activity
          protein heterodimerization activity
          actin filament binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0