Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PPM1A SIRNA Assays ›
          Gene Symbol:
          PPM1A
          Gene Name:
          protein phosphatase, Mg2+/Mn2+ dependent 1A
          Gene Aliases:
          PP2C-ALPHA, PP2CA, PP2Calpha
          Chromosome Location:
          Chr.14: 60245752 - 60299087 on Build GRCh38
          UniGene ID:
          Hs.130036
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 42904

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 286.00
          Your Price
          Online offer: 277.65
          277.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PPM1A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_021003.4 NP_066283.1 2 549
          NM_177951.2 NP_808820.1 3 651
          NM_177952.2 NP_808821.2 2 414
          XM_005267779.1 XP_005267836.1 3 496
          XM_005267781.1 XP_005267838.1 3 654
          XM_011536872.1 XP_011535174.1 5 916
          XM_011536876.1 XP_011535178.1 4 773
          XM_011536877.1 XP_011535179.1 5 816
          XM_011536878.1 XP_011535180.1 4 739
          XM_011536879.2 XP_011535181.1 4 608
          XM_011536880.2 XP_011535182.1 5 662
          XM_011536881.2 XP_011535183.1 3 522
          XM_011536882.2 XP_011535184.1 2 191
          XM_011536883.1 XP_011535185.1 3 605
          XM_011536884.1 XP_011535186.1 2 162
          XM_017021381.1 XP_016876870.1 5 1786
          XM_017021382.1 XP_016876871.1 7 1036
          XM_017021383.1 XP_016876872.1 4 672
          XM_017021384.1 XP_016876873.1 4 364
          XM_017021385.1 XP_016876874.1 3 639
          XM_017021386.1 XP_016876875.1 3 402
          XM_017021387.1 XP_016876876.1 4 648
          GenBank mRNA AB451247.1 - 1 99
          AB451371.1 - 1 99
          AF070670.1 - 3 458
          AK097843.1 - 2 414
          AK308225.1 - 2 278
          AK313337.1 - 3 496
          AY236965.1 - 2 179
          BC026691.1 - 2 402 3847638
          BC063243.1 - 2 332 6011486
          DA439296.1 - 3 451
          S87759.1 - 2 456

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          protein dephosphorylation
          N-terminal protein myristoylation
          cell cycle arrest
          negative regulation of SMAD protein complex assembly
          Wnt signaling pathway
          dephosphorylation
          positive regulation of Wnt signaling pathway
          negative regulation of transforming growth factor beta receptor signaling pathway
          negative regulation of BMP signaling pathway
          peptidyl-threonine dephosphorylation
          negative regulation of NF-kappaB import into nucleus
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of transcription, DNA-templated
          positive regulation of protein export from nucleus
          cellular response to transforming growth factor beta stimulus
          magnesium ion binding
          protein serine/threonine phosphatase activity
          signal transducer activity
          protein binding
          manganese ion binding
          calmodulin-dependent protein phosphatase activity
          R-SMAD binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0