Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYB561 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017916.1 | NP_001017916.1 | 6 | 2210 | ||
NM_001017917.1 | NP_001017917.1 | 6 | 2043 | |||
NM_001915.3 | NP_001906.3 | 6 | 2040 | |||
XM_005257091.1 | XP_005257148.1 | 6 | 1985 | |||
XM_017024260.1 | XP_016879749.1 | 6 | 2082 | |||
XM_017024261.1 | XP_016879750.1 | 5 | 1949 | |||
XM_017024262.1 | XP_016879751.1 | 6 | 2275 | |||
GenBank mRNA | AK095244.1 | - | 6 | 2257 | ||
BC002976.1 | - | 6 | 1997 | 3535771 | ||
BC091485.1 | - | 6 | 2011 | 6188259 |