Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HOXB3 SIRNA Assays ›
Gene Symbol:
HOXB3
Gene Name:
homeobox B3
Gene Aliases:
HOX2, HOX2G, Hox-2.7
Chromosome Location:
Chr.17: 48548870 - 48590369 on Build GRCh38
UniGene ID:
Hs.654560
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
4298
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HOXB3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_002146.4
NP_002137.4
4
2123
XM_005257277.3
XP_005257334.1
5
1837
XM_005257280.3
XP_005257337.1
5
1748
XM_005257282.3
XP_005257339.1
5
1313
XM_006721854.2
XP_006721917.1
5
1855
XM_011524707.2
XP_011523009.1
5
11663
XM_011524708.2
XP_011523010.1
4
11243
XM_011524710.1
XP_011523012.1
5
4063
XM_011524719.1
XP_011523021.1
4
6544
XM_011524720.2
XP_011523022.1
4
9235
XM_011524721.2
XP_011523023.1
4
4367
XM_011524726.2
XP_011523028.1
4
1321
XM_017024557.1
XP_016880046.1
5
11411
XM_017024558.1
XP_016880047.1
5
11285
XM_017024559.1
XP_016880048.1
5
11127
XM_017024560.1
XP_016880049.1
5
11371
XM_017024561.1
XP_016880050.1
5
15208
XM_017024562.1
XP_016880051.1
5
11149
XM_017024563.1
XP_016880052.1
6
13804
GenBank mRNA
AK027609.1
-
4
2161
AK291182.1
-
5
1834
AK299226.1
-
5
1181
AK299265.1
-
5
1580
AK309721.1
-
2
1001
AK315102.1
-
3
1389
AK316245.1
-
4
1356
U59298.1
-
2
2714
X16667.1
-
4
1628
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001107042, XM_006247194, BC166859.
This siRNA also targets the following transcripts in Mouse: NM_010458, NM_001079869, S35628, S35738, X66177.
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
angiogenesis
hematopoietic progenitor cell differentiation
transcription, DNA-templated
anterior/posterior pattern specification
rhombomere development
glossopharyngeal nerve morphogenesis
thyroid gland development
embryonic skeletal system morphogenesis
regulation of neurogenesis
cartilage development
definitive hemopoiesis
face development
RNA polymerase II core promoter sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0