Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RUNX1T1 SIRNA Assays ›
Gene Symbol:
RUNX1T1
Gene Name:
RUNX1 translocation partner 1
Gene Aliases:
AML1-MTG8, AML1T1, CBFA2T1, CDR, ETO, MTG8, ZMYND2
Chromosome Location:
Chr.8: 91954967 - 92103365 on Build GRCh38
UniGene ID:
Hs.368431
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
43150
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RUNX1T1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001198625.1
NP_001185554.1
6
786
NM_001198626.1
NP_001185555.1
4
644
NM_001198627.1
NP_001185556.1
5
704
NM_001198628.1
NP_001185557.1
3
301
NM_001198629.1
NP_001185558.1
3
427
NM_001198630.1
NP_001185559.1
4
487
NM_001198631.1
NP_001185560.1
3
500
NM_001198632.1
NP_001185561.1
3
418
NM_001198633.1
NP_001185562.1
2
389
NM_001198634.1
NP_001185563.1
2
327
NM_001198679.1
NP_001185608.1
3
471
NM_004349.3
NP_004340.1
2
437
NM_175634.2
NP_783552.1
3
562
NM_175635.2
NP_783553.1
2
254
NM_175636.2
NP_783554.1
1
336
XM_006716676.3
XP_006716739.1
4
371
XM_011517351.2
XP_011515653.1
4
416
XM_011517352.2
XP_011515654.1
4
656
XM_011517353.2
XP_011515655.1
3
364
XM_017013930.1
XP_016869419.1
3
672
XM_017013931.1
XP_016869420.1
2
194
XM_017013932.1
XP_016869421.1
4
559
XM_017013933.1
XP_016869422.1
5
623
XM_017013934.1
XP_016869423.1
5
731
XM_017013935.1
XP_016869424.1
4
536
XM_017013936.1
XP_016869425.1
3
630
XM_017013937.1
XP_016869426.1
4
456
XM_017013938.1
XP_016869427.1
3
320
GenBank mRNA
AA506749.1
-
1
343
AK297616.1
-
2
151
AK312592.1
-
6
548
BC005850.1
-
2
216
2961112
BC067078.1
-
2
207
2964656
CR456792.1
-
1
26
D14289.1
-
1
293
D14821.1
-
2
191
D43638.1
-
2
437
DA499596.1
-
4
417
DA500281.1
-
3
391
DA504975.1
-
4
524
DA770917.1
-
2
254
DA774453.1
-
3
400
DA776825.1
-
3
320
DA776968.1
-
4
499
DA777240.1
-
3
386
DA779230.1
-
2
350
DA795825.1
-
5
520
DA796892.1
-
3
301
KF946505.1
-
2
55
KF946506.1
-
2
55
KF946507.1
-
2
55
KF946508.1
-
2
55
KF946509.1
-
2
55
KF946510.1
-
2
55
KF946511.1
-
2
41
KF946512.1
-
3
1345
KF946513.1
-
2
1725
KF946514.1
-
2
41
KF946515.1
-
1
281
KF946516.1
-
1
281
KF946517.1
-
1
281
KF946518.1
-
1
281
X79990.1
-
1
19
Z35296.1
-
204
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001111026, NM_001111027, NM_009822, AK049615, AK032132, and others.
Panther Classification:
Molecular Function -
transcription cofactor
Gene Ontology Categories:
Function(s)
Process(es)
generation of precursor metabolites and energy
transcription, DNA-templated
negative regulation of fat cell differentiation
negative regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription corepressor activity
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0