Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CD86 SIRNA Assays ›
          Gene Symbol:
          CD86
          Gene Name:
          CD86 molecule
          Gene Aliases:
          B7-2, B7.2, B70, CD28LG2, LAB72
          Chromosome Location:
          Chr.3: 122055362 - 122121143 on Build GRCh38
          UniGene ID:
          Hs.171182
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 43161

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CD86
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001206925.1 NP_001193854.1 2 174
          NM_006889.4 NP_008820.3 3 279
          NM_175862.4 NP_787058.4 3 224
          NM_176892.1 NP_795711.1 3 279
          GenBank mRNA AK291497.1 - 3 132
          AK313103.1 - 3 133
          AK316203.1 - 2 170
          BC040261.1 - 3 212 5173789
          CD705810.1 - 3 301
          CR541844.1 - 2 78
          DA619360.1 - 3 225
          KU284848.1 - 2 78
          L25259.1 - 3 202
          U04343.1 - 3 225

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          immunoglobulin receptor superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          response to yeast
          adaptive immune response
          T cell proliferation involved in immune response
          negative regulation of T cell anergy
          immune response
          cell-cell signaling
          aging
          positive regulation of cell proliferation
          regulation of phosphatidylinositol 3-kinase signaling
          T cell costimulation
          toll-like receptor 3 signaling pathway
          response to interferon-gamma
          positive regulation of activated T cell proliferation
          T cell activation
          B cell activation
          response to drug
          myeloid dendritic cell differentiation
          positive regulation of lymphotoxin A biosynthetic process
          positive regulation of interleukin-2 biosynthetic process
          positive regulation of interleukin-4 biosynthetic process
          positive regulation of T-helper 2 cell differentiation
          positive regulation of transcription, DNA-templated
          viral entry into host cell
          phosphatidylinositol phosphorylation
          phosphatidylinositol-mediated signaling
          defense response to virus
          cellular response to lipopolysaccharide
          cellular response to metal ion
          cellular response to cytokine stimulus
          virus receptor activity
          receptor activity
          receptor binding
          protein binding
          coreceptor activity
          phosphatidylinositol-4,5-bisphosphate 3-kinase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0