Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RUNX1T1 SIRNA Assays ›
          Gene Symbol:
          RUNX1T1
          Gene Name:
          RUNX1 translocation partner 1
          Gene Aliases:
          AML1-MTG8, AML1T1, CBFA2T1, CDR, ETO, MTG8, ZMYND2
          Chromosome Location:
          Chr.8: 91954967 - 92103365 on Build GRCh38
          UniGene ID:
          Hs.368431
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 43218

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RUNX1T1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001198625.1 NP_001185554.1 6 805
          NM_001198626.1 NP_001185555.1 4 663
          NM_001198627.1 NP_001185556.1 5 723
          NM_001198628.1 NP_001185557.1 3 320
          NM_001198629.1 NP_001185558.1 3 446
          NM_001198630.1 NP_001185559.1 4 506
          NM_001198631.1 NP_001185560.1 3 519
          NM_001198632.1 NP_001185561.1 3 437
          NM_001198633.1 NP_001185562.1 2 408
          NM_001198634.1 NP_001185563.1 2 346
          NM_001198679.1 NP_001185608.1 3 490
          NM_004349.3 NP_004340.1 2 456
          NM_175634.2 NP_783552.1 3 581
          NM_175635.2 NP_783553.1 2 273
          NM_175636.2 NP_783554.1 1 355
          XM_006716676.3 XP_006716739.1 4 390
          XM_011517351.2 XP_011515653.1 4 435
          XM_011517352.2 XP_011515654.1 4 675
          XM_011517353.2 XP_011515655.1 3 383
          XM_017013930.1 XP_016869419.1 3 691
          XM_017013931.1 XP_016869420.1 2 213
          XM_017013932.1 XP_016869421.1 4 578
          XM_017013933.1 XP_016869422.1 5 642
          XM_017013934.1 XP_016869423.1 5 750
          XM_017013935.1 XP_016869424.1 4 555
          XM_017013936.1 XP_016869425.1 3 649
          XM_017013937.1 XP_016869426.1 4 475
          XM_017013938.1 XP_016869427.1 3 339
          GenBank mRNA AA506749.1 - 1 362
          AK297616.1 - 2 170
          AK312592.1 - 6 567
          BC005850.1 - 2 235 2961112
          BC067078.1 - 2 226 2964656
          BT009871.1 - 1 15
          CR456792.1 - 1 45
          D14289.1 - 1 312
          D14821.1 - 2 210
          D43638.1 - 2 456
          DA499596.1 - 4 436
          DA500281.1 - 3 410
          DA770917.1 - 2 273
          DA774453.1 - 3 419
          DA776825.1 - 3 339
          DA776968.1 - 4 518
          DA777240.1 - 3 405
          DA779230.1 - 2 369
          DA796892.1 - 3 320
          KF946505.1 - 2 74
          KF946506.1 - 2 74
          KF946507.1 - 2 74
          KF946508.1 - 2 74
          KF946509.1 - 2 74
          KF946510.1 - 2 74
          KF946511.1 - 2 60
          KF946512.1 - 3 1364
          KF946513.1 - 2 1744
          KF946514.1 - 2 60
          KF946515.1 - 1 300
          KF946516.1 - 1 300
          KF946517.1 - 1 300
          KF946518.1 - 1 300
          X79990.1 - 1 38
          Z35296.1 - 223

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001108657, XM_006237903, XM_008763549, XM_006237904, XM_008763550, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001111026, NM_009822, NM_001111027, AK049615, AK032132, and others.

          Panther Classification:

          Molecular Function -

          transcription cofactor

          Gene Ontology Categories:

          Function(s) Process(es)

          generation of precursor metabolites and energy
          transcription, DNA-templated
          negative regulation of fat cell differentiation
          negative regulation of transcription, DNA-templated
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          transcription corepressor activity
          protein binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0