Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANKRD11 SIRNA Assays ›
Gene Symbol:
ANKRD11
Gene Name:
ankyrin repeat domain 11
Gene Aliases:
ANCO-1, ANCO1, LZ16, T13
Chromosome Location:
Chr.16: 89267619 - 89490561 on Build GRCh38
UniGene ID:
Hs.335003
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
43376
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANKRD11
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256182.1
NP_001243111.1
4
609
NM_001256183.1
NP_001243112.1
3
535
NM_013275.5
NP_037407.4
3
538
NR_045839.1
3
538
XM_006721181.1
XP_006721244.1
2
218
XM_006721184.2
XP_006721247.1
1
80
XM_011523051.2
XP_011521353.1
4
598
XM_011523053.2
XP_011521355.1
4
704
XM_011523054.2
XP_011521356.1
3
521
XM_011523055.2
XP_011521357.1
3
533
XM_011523056.2
XP_011521358.1
3
521
XM_011523057.2
XP_011521359.1
3
521
XM_017023182.1
XP_016878671.1
3
971
XM_017023183.1
XP_016878672.1
4
707
XM_017023184.1
XP_016878673.1
4
704
XM_017023185.1
XP_016878674.1
2
345
XM_017023186.1
XP_016878675.1
6
1081
XM_017023187.1
XP_016878676.1
2
433
GenBank mRNA
AB209186.1
-
2
183
AF121775.1
-
3
295
AK125549.1
-
2
514
AK290994.1
-
2
217
AY373756.1
-
3
538
AY533563.1
-
4
366
BC017437.1
-
3
507
BC025283.1
-
2
162
BC052290.1
-
3
453
BC069013.1
-
3
453
BQ957747.1
-
526
KJ534770.1
-
1
77
KM670019.1
-
136
KM670020.1
-
136
Back To Top
More Information
Gene Ontology Categories:
Function(s)
in utero embryonic development
tissue homeostasis
multicellular organism growth
odontogenesis of dentin-containing tooth
skeletal system morphogenesis
face morphogenesis
bone development
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-9fjjm:80/10.239.165.64:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0