Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ERC1 SIRNA Assays ›
          Gene Symbol:
          ERC1
          Gene Name:
          ELKS/RAB6-interacting/CAST family member 1
          Gene Aliases:
          Cast2, ELKS, ERC-1, RAB6IP2
          Chromosome Location:
          Chr.12: 990010 - 1495933 on Build GRCh38
          UniGene ID:
          Hs.601216
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 43397

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ERC1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001301248.1 NP_001288177.1 1 313
          NM_178039.3 NP_829883.1 2 428
          NM_178040.3 NP_829884.1 2 428
          NR_027946.2 2 593
          NR_027948.2 2 593
          NR_027949.2 2 428
          XM_011520941.2 XP_011519243.1 2 472
          XM_017019052.1 XP_016874541.1 2 538
          XM_017019053.1 XP_016874542.1 2 472
          XM_017019054.1 XP_016874543.1 2 398
          XM_017019055.1 XP_016874544.1 2 394
          XM_017019056.1 XP_016874545.1 2 335
          XM_017019057.1 XP_016874546.1 2 538
          XM_017019058.1 XP_016874547.1 2 472
          XM_017019059.1 XP_016874548.1 2 538
          XM_017019060.1 XP_016874549.1 2 538
          XM_017019061.1 XP_016874550.1 2 538
          XM_017019062.1 XP_016874551.1 2 538
          XM_017019063.1 XP_016874552.1 2 538
          XM_017019064.1 XP_016874553.1 2 538
          XM_017019065.1 XP_016874554.1 2 538
          XM_017019066.1 XP_016874555.1 2 538
          XM_017019067.1 XP_016874556.1 2 538
          XM_017019068.1 XP_016874557.1 2 538
          XM_017019069.1 XP_016874558.1 2 538
          XM_017019070.1 XP_016874559.1 2 538
          XM_017019071.1 XP_016874560.1 2 538
          XM_017019072.1 XP_016874561.1 2 538
          XM_017019073.1 XP_016874562.1 2 538
          GenBank mRNA AB015617.1 - 2 551
          AB029004.2 - 1 280
          AB053468.1 - 2 534
          AB053469.1 - 2 363
          AB053470.1 - 2 370
          AB053471.1 - 2 338
          AK091577.1 - 2 538
          BC068006.1 - 1 279
          BC132782.1 - 1 289 40146804
          BC132784.1 - 1 289 40146806
          BC144287.1 - 1 289
          BC150248.1 - 1 301 9007473
          DR000847.1 - 2 451

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_170788, XM_006237230, XM_006237226, XM_006237227, XM_008763221, and others.
          This siRNA also targets the following transcripts in Mouse: NM_053204, AK053824, AY316692, AK035989, AK134383, and others.

          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          I-kappaB phosphorylation
          multicellular organism development
          protein transport
          retrograde transport, endosome to Golgi
          positive regulation of NF-kappaB transcription factor activity
          cell-cell adhesion
          protein binding
          Rab GTPase binding
          PDZ domain binding
          leucine zipper domain binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0