Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTMR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016156.5 | NP_057240.3 | 1, 2 | 341 | ||
XM_005274375.2 | XP_005274432.1 | 1, 2 | 244 | |||
XM_006718936.3 | XP_006718999.1 | 1, 2 | 306 | |||
XM_017018517.1 | XP_016874006.1 | 1, 2 | 278 | |||
GenBank mRNA | AB028996.1 | - | 1, 2 | 429 | ||
AK291277.1 | - | 1, 2 | 115 | |||
BC040432.1 | - | 1, 2 | 93 | |||
BC052990.1 | - | 1, 2 | 226 | 6501680 | ||
BM475570.1 | - | 1, 2 | 226 | |||
BP358035.1 | - | 1, 2 | 341 |