Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRODH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016335.4 | NP_057419.4 | 2, 3 | 482 | ||
GenBank mRNA | AB209472.1 | - | 1, 2 | 133 | ||
BC036534.1 | - | 1, 2 | 291 | |||
BC068260.1 | - | 1, 2 | 284 | |||
CN362765.1 | - | 1, 2 | 233 | |||
KJ534920.1 | - | 1, 2 | 278 | |||
U79754.1 | - | 2, 3 | 482 |
Gene Symbol: LOC102724788 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006724935.3 | XP_006724998.1 | 1, 2 | 311 | ||
XM_006724936.3 | XP_006724999.1 | 1, 2 | 286 |