Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FAM65B SIRNA Assays ›
          Gene Symbol:
          FAM65B
          Gene Name:
          family with sequence similarity 65 member B
          Gene Aliases:
          C6orf32, DFNB104, DIFF40, DIFF48, MYONAP, PL48
          Chromosome Location:
          Chr.6: 24804281 - 25042288 on Build GRCh38
          UniGene ID:
          Hs.559459
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 43499

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FAM65B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001286445.1 NP_001273374.1 2, 3 350
          NM_001286446.1 NP_001273375.1 2, 3 453
          NM_001286447.1 NP_001273376.1 2, 3 550
          NM_014722.3 NP_055537.2 2, 3 488
          NM_015864.3 NP_056948.2 2, 3 550
          XM_006715275.2 XP_006715338.1 2, 3 567
          XM_006715277.1 XP_006715340.1 2, 3 488
          XM_006715279.2 XP_006715342.1 2, 3 545
          XM_006715281.3 XP_006715344.1 2, 3 567
          XM_011515007.1 XP_011513309.1 3, 4 1076
          XM_011515008.2 XP_011513310.1 3, 4 407
          XM_011515009.1 XP_011513311.1 2, 3 279
          XM_011515010.2 XP_011513312.1 2, 3 320
          XM_011515012.1 XP_011513314.1 2, 3 567
          XM_017011523.1 XP_016867012.1 2, 3 242
          XM_017011524.1 XP_016867013.1 2, 3 496
          XM_017011525.1 XP_016867014.1 2, 3 488
          XM_017011526.1 XP_016867015.1 2, 3 488
          XM_017011527.1 XP_016867016.1 2, 3 320
          GenBank mRNA AB002384.1 - 2, 3 286
          AK299047.1 - 2, 3 258
          AK300138.1 - 2, 3 498
          AK300950.1 - 2, 3 303
          BC001232.1 - 2, 3 540 3453890
          CN313643.1 - 2, 3 550
          DA258515.1 - 2, 3 352
          DC406827.1 - 2, 3 454
          DC428807.1 - 2, 3 350
          U49187.1 - 2, 3 412

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: XM_006253930, XM_008771639, XM_008771637, XM_008771638, XM_006253929, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001080381, NM_029679, NM_001286101, NM_001286100, NM_178658, and others.

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          sensory perception of sound
          skeletal muscle fiber development
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0