Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP3A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017460.5 | NP_059488.2 | 7, 8 | 788 | ||
XM_017011780.1 | XP_016867269.1 | 7, 8 | 786 | |||
GenBank mRNA | AF182273.1 | - | 7, 8 | 690 | ||
AK298451.1 | - | 6, 7 | 659 | |||
AK298462.1 | - | 5, 6 | 535 | |||
AK312967.1 | - | 7, 8 | 781 | |||
AY606313.2 | - | 7, 8 | 683 | |||
BC069418.1 | - | 7, 8 | 731 | 7262313 | ||
BC101631.1 | - | 7, 8 | 777 | 8069137 | ||
D00003.1 | - | 7, 8 | 741 | |||
DQ924960.1 | - | 7, 8 | 681 | |||
M13785.1 | - | 7, 8 | 741 | |||
M14096.1 | - | 7, 8 | 749 | |||
M18907.1 | - | 7, 8 | 771 | |||
X12387.1 | - | 7, 8 | 750 |