Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANKRD36 SIRNA Assays ›
Gene Symbol:
ANKRD36
ANKRD36B
ANKRD36C
Gene Name:
ankyrin repeat domain 36
ankyrin repeat domain 36B
ankyrin repeat domain 36C
Gene Aliases:
UNQ2430
KIAA1641
-
Chromosome Location:
Chr.2: 97113186 - 97264521 on Build GRCh38
Chromosome Location:
Chr.2: 97492663 - 97590245 on Build GRCh38
Chromosome Location:
Chr.2: 95849548 - 95992320 on Build GRCh38
UniGene ID:
Hs.541894
Hs.532921
Hs.646318
Species:
Human
Species Specific ID (Flybase ID):
-
-
-
siRNA ID
43631
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANKRD36
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001164315.1
NP_001157787.1
71, 72
5734
XM_006712514.3
XP_006712577.1
71, 72
5966
XM_011511130.1
XP_011509432.1
71, 72
5966
XM_017004009.1
XP_016859498.1
71, 72
5966
XM_017004010.1
XP_016859499.1
71, 72
5963
XM_017004011.1
XP_016859500.1
69, 70
5864
XM_017004012.1
XP_016859501.1
69, 70
5864
XM_017004013.1
XP_016859502.1
69, 70
5864
XM_017004014.1
XP_016859503.1
69, 70
5864
XM_017004015.1
XP_016859504.1
69, 70
5864
XM_017004016.1
XP_016859505.1
69, 70
5864
XM_017004017.1
XP_016859506.1
69, 70
5864
XM_017004018.1
XP_016859507.1
69, 70
5864
XM_017004019.1
XP_016859508.1
69, 70
5864
XM_017004020.1
XP_016859509.1
69, 70
5864
XM_017004021.1
XP_016859510.1
69, 70
5864
XM_017004022.1
XP_016859511.1
69, 70
5864
XM_017004023.1
XP_016859512.1
69, 70
5864
XM_017004024.1
XP_016859513.1
70, 71
5828
XM_017004025.1
XP_016859514.1
67, 68
5762
XM_017004026.1
XP_016859515.1
67, 68
5762
XM_017004027.1
XP_016859516.1
67, 68
5762
XM_017004028.1
XP_016859517.1
67, 68
5762
XM_017004029.1
XP_016859518.1
67, 68
5762
XM_017004030.1
XP_016859519.1
67, 68
5762
XM_017004031.1
XP_016859520.1
67, 68
5762
XM_017004032.1
XP_016859521.1
67, 68
5762
XM_017004033.1
XP_016859522.1
67, 68
5762
XM_017004034.1
XP_016859523.1
67, 68
5762
XM_017004035.1
XP_016859524.1
67, 68
5762
XM_017004036.1
XP_016859525.1
67, 68
5762
XM_017004037.1
XP_016859526.1
67, 68
5762
XM_017004038.1
XP_016859527.1
67, 68
5762
XM_017004039.1
XP_016859528.1
65, 66
5660
XM_017004040.1
XP_016859529.1
65, 66
5660
XM_017004041.1
XP_016859530.1
65, 66
5660
XM_017004042.1
XP_016859531.1
65, 66
5660
XM_017004043.1
XP_016859532.1
65, 66
5660
XM_017004044.1
XP_016859533.1
63, 64
5558
XM_017004045.1
XP_016859534.1
63, 64
5558
XM_017004046.1
XP_016859535.1
69, 70
5721
XM_017004047.1
XP_016859536.1
68, 69
5653
XM_017004048.1
XP_016859537.1
67, 68
5580
XM_017004049.1
XP_016859538.1
69, 70
5755
XM_017004050.1
XP_016859539.1
54, 55
4159
XM_017004051.1
XP_016859540.1
52, 53
4373
GenBank mRNA
AB046861.1
-
13, 14
2632
AF543495.1
-
3, 4
1175
AK090926.1
-
1, 2
826
BC146835.1
-
13, 14
2643
8860491
Gene Symbol: ANKRD36B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_025190.3
NP_079466.3
39, 40
4006
XM_005263995.4
XP_005264052.1
39, 40
4286
XM_006712661.1
XP_006712724.1
39, 40
4286
XM_017004578.1
XP_016860067.1
39, 40
4283
XM_017004579.1
XP_016860068.1
37, 38
4184
XM_017004580.1
XP_016860069.1
37, 38
4184
XM_017004581.1
XP_016860070.1
37, 38
4184
XM_017004582.1
XP_016860071.1
37, 38
4184
XM_017004583.1
XP_016860072.1
38, 39
4148
XM_017004584.1
XP_016860073.1
33, 34
3980
XM_017004585.1
XP_016860074.1
37, 38
4041
XM_017004586.1
XP_016860075.1
34, 35
3027
XM_017004587.1
XP_016860076.1
38, 39
4215
XM_017004588.1
XP_016860077.1
37, 38
4080
XM_017004589.1
XP_016860078.1
36, 37
3973
XM_017004590.1
XP_016860079.1
35, 36
3900
XM_017004591.1
XP_016860080.1
21, 22
4004
XM_017004592.1
XP_016860081.1
40, 41
4330
XM_017004593.1
XP_016860082.1
38, 39
4136
XM_017004594.1
XP_016860083.1
37, 38
4029
XM_017004595.1
XP_016860084.1
38, 39
4085
GenBank mRNA
AF432208.1
-
1, 2
376
AF432209.1
-
1, 2
473
AK024934.1
-
1, 2
118
AK056042.1
-
1, 2
235
BC038460.1
-
1, 2
192
BC125132.1
-
1, 2
118
BC125133.1
-
1, 2
118
DB105321.1
-
1, 2
165
Gene Symbol: ANKRD36C
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001310154.1
NP_001297083.1
84, 85
6711
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
ion channel inhibitor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0