Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC38A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001307936.1 | NP_001294865.1 | 11 | 1300 | ||
NM_018976.4 | NP_061849.2 | 12 | 1416 | |||
GenBank mRNA | AB037803.2 | - | 11 | 976 | ||
AF259799.1 | - | 11 | 996 | |||
AF298897.1 | - | 12 | 1327 | |||
AJ344099.1 | - | 12 | 1328 | |||
AK001700.1 | - | 11 | 1209 | |||
AK172784.1 | - | 5 | 423 | |||
AK291644.1 | - | 12 | 1120 | |||
AK293858.1 | - | 5 | 531 | |||
BC040342.1 | - | 12 | 1318 | 3874551 | ||
CR457267.1 | - | 8 | 676 |