Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other AGPAT3 SIRNA Assays ›
          Gene Symbol:
          AGPAT3
          Gene Name:
          1-acylglycerol-3-phosphate O-acyltransferase 3
          Gene Aliases:
          LPAAT-GAMMA1, LPAAT3
          Chromosome Location:
          Chr.21: 43865160 - 43987594 on Build GRCh38
          UniGene ID:
          Hs.248785
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 43765

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: AGPAT3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001037553.1 NP_001032642.1 9 1182
          NM_020132.4 NP_064517.1 10 1302
          XM_005261160.4 XP_005261217.1 10 1303
          XM_006724029.3 XP_006724092.1 9 1231
          XM_006724030.3 XP_006724093.1 9 1199
          XM_006724031.3 XP_006724094.1 9 1202
          XM_011529660.2 XP_011527962.1 9 1041
          XM_011529661.2 XP_011527963.1 9 1356
          XM_011529662.2 XP_011527964.1 11 2006
          XM_011529663.2 XP_011527965.1 9 1215
          XM_011529664.2 XP_011527966.1 9 1156
          XM_011529665.2 XP_011527967.1 11 1657
          XM_017028408.1 XP_016883897.1 9 1185
          XM_017028409.1 XP_016883898.1 9 1263
          XM_017028410.1 XP_016883899.1 9 1268
          XM_017028411.1 XP_016883900.1 9 1141
          XM_017028412.1 XP_016883901.1 10 2234
          XM_017028413.1 XP_016883902.1 10 1202
          GenBank mRNA AB040138.1 - 9 1257
          AF156774.1 - 10 1263
          AF156775.1 - 9 1126
          AK125804.1 - 7 3919
          AL832919.1 - 7 875
          BC011971.1 - 10 1276 4148915
          BC040603.1 - 11 2006 5270905
          BC063552.1 - 9 1572 4800514
          BM476336.1 - 2 158

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          acyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          phosphatidic acid biosynthetic process
          phospholipid biosynthetic process
          CDP-diacylglycerol biosynthetic process
          triglyceride biosynthetic process
          1-acylglycerol-3-phosphate O-acyltransferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-6hb64:80/100.66.77.245:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0