Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANKRD36B SIRNA Assays ›
Gene Symbol:
ANKRD36B
ANKRD36
Gene Name:
ankyrin repeat domain 36B
ankyrin repeat domain 36
Gene Aliases:
KIAA1641
UNQ2430
Chromosome Location:
Chr.2: 97492663 - 97590245 on Build GRCh38
Chromosome Location:
Chr.2: 97113186 - 97264521 on Build GRCh38
UniGene ID:
Hs.532921
Hs.541894
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
43774
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANKRD36B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_025190.3
NP_079466.3
40
4039
XM_005263995.4
XP_005264052.1
40
4319
XM_006712661.1
XP_006712724.1
40
4319
XM_017004578.1
XP_016860067.1
40
4316
XM_017004579.1
XP_016860068.1
38
4217
XM_017004580.1
XP_016860069.1
38
4217
XM_017004581.1
XP_016860070.1
38
4217
XM_017004582.1
XP_016860071.1
38
4217
XM_017004583.1
XP_016860072.1
39
4181
XM_017004584.1
XP_016860073.1
34
4013
XM_017004585.1
XP_016860074.1
38
4074
XM_017004586.1
XP_016860075.1
35
3060
XM_017004587.1
XP_016860076.1
39
4248
XM_017004588.1
XP_016860077.1
38
4113
XM_017004589.1
XP_016860078.1
37
4006
XM_017004590.1
XP_016860079.1
36
3933
XM_017004591.1
XP_016860080.1
22
4037
XM_017004592.1
XP_016860081.1
41
4363
XM_017004593.1
XP_016860082.1
39
4169
XM_017004594.1
XP_016860083.1
38
4062
XM_017004595.1
XP_016860084.1
39
4118
GenBank mRNA
AF432208.1
-
2
409
AF432209.1
-
2
506
AK024934.1
-
2
151
AK056042.1
-
2
268
BC038460.1
-
2
225
BC125132.1
-
2
151
BC125133.1
-
2
151
DB105321.1
-
2
198
Gene Symbol: ANKRD36
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001164315.1
NP_001157787.1
72
5767
XM_006712514.3
XP_006712577.1
72
5999
XM_011511130.1
XP_011509432.1
72
5999
XM_017004009.1
XP_016859498.1
72
5999
XM_017004010.1
XP_016859499.1
72
5996
XM_017004011.1
XP_016859500.1
70
5897
XM_017004012.1
XP_016859501.1
70
5897
XM_017004013.1
XP_016859502.1
70
5897
XM_017004014.1
XP_016859503.1
70
5897
XM_017004015.1
XP_016859504.1
70
5897
XM_017004016.1
XP_016859505.1
70
5897
XM_017004017.1
XP_016859506.1
70
5897
XM_017004018.1
XP_016859507.1
70
5897
XM_017004019.1
XP_016859508.1
70
5897
XM_017004020.1
XP_016859509.1
70
5897
XM_017004021.1
XP_016859510.1
70
5897
XM_017004022.1
XP_016859511.1
70
5897
XM_017004023.1
XP_016859512.1
70
5897
XM_017004024.1
XP_016859513.1
71
5861
XM_017004025.1
XP_016859514.1
68
5795
XM_017004026.1
XP_016859515.1
68
5795
XM_017004027.1
XP_016859516.1
68
5795
XM_017004028.1
XP_016859517.1
68
5795
XM_017004029.1
XP_016859518.1
68
5795
XM_017004030.1
XP_016859519.1
68
5795
XM_017004031.1
XP_016859520.1
68
5795
XM_017004032.1
XP_016859521.1
68
5795
XM_017004033.1
XP_016859522.1
68
5795
XM_017004034.1
XP_016859523.1
68
5795
XM_017004035.1
XP_016859524.1
68
5795
XM_017004036.1
XP_016859525.1
68
5795
XM_017004037.1
XP_016859526.1
68
5795
XM_017004038.1
XP_016859527.1
68
5795
XM_017004039.1
XP_016859528.1
66
5693
XM_017004040.1
XP_016859529.1
66
5693
XM_017004041.1
XP_016859530.1
66
5693
XM_017004042.1
XP_016859531.1
66
5693
XM_017004043.1
XP_016859532.1
66
5693
XM_017004044.1
XP_016859533.1
64
5591
XM_017004045.1
XP_016859534.1
64
5591
XM_017004046.1
XP_016859535.1
70
5754
XM_017004047.1
XP_016859536.1
69
5686
XM_017004048.1
XP_016859537.1
68
5613
XM_017004049.1
XP_016859538.1
70
5788
XM_017004050.1
XP_016859539.1
55
4192
XM_017004051.1
XP_016859540.1
53
4406
GenBank mRNA
AB046861.1
-
14
2665
AF543495.1
-
4
1208
AK090926.1
-
2
859
BC146835.1
-
14
2676
8860491
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0