Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TPM3 SIRNA Assays ›
          Gene Symbol:
          TPM3
          Gene Name:
          tropomyosin 3
          Gene Aliases:
          CAPM1, CFTD, HEL-189, HEL-S-82p, NEM1, OK/SW-cl.5, TM-5, TM3, TM30, TM30nm, TM5, TPMsk3, TRK, hscp30
          Chromosome Location:
          Chr.1: 154155304 - 154192135 on Build GRCh38
          UniGene ID:
          Hs.535581
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 43886

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TPM3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001043351.1 NP_001036816.1 2, 3 406
          NM_001043352.1 NP_001036817.1 2, 3 406
          NM_001043353.1 NP_001036818.1 2, 3 406
          NM_001278188.1 NP_001265117.1 1, 2 272
          NM_001278189.1 NP_001265118.1 2, 3 406
          NM_001278190.1 NP_001265119.1 2, 3 406
          NM_001278191.1 NP_001265120.1 2, 3 252
          NM_152263.3 NP_689476.2 3, 4 504
          NM_153649.3 NP_705935.1 2, 3 406
          NR_103461.1 2, 3 406
          GenBank mRNA AF474157.1 - 2, 3 276
          AK026559.1 - 2, 3 346
          AK056921.1 - 2, 3 263
          AK056997.1 - 3, 4 294
          AK092690.1 - 3, 4 344
          AK125367.1 - 1, 2 170
          AK298678.1 - 1, 2 250
          AK301674.1 - 2, 3 228
          AK315504.1 - 2, 3 365
          AL537561.3 - 2, 3 316
          AY004867.1 - 2, 3 287
          BC000771.2 - 2, 3 318 3506357
          BC008407.1 - 3, 4 470 4245271
          BC008425.1 - 3, 4 473 4246114
          BC015403.1 - 2, 3 321 4389911
          BC062740.1 - 3, 4 336 4271142
          BC072428.1 - 2, 3 333 6472532
          BF967161.1 - 2, 3 219
          BG706843.1 - 2, 3 349
          BQ933569.1 - 2, 3 373
          BU956481.1 - 2, 3 332
          BX419658.2 - 2, 3 319
          CR997359.1 - 2, 3 297
          DA209890.1 - 2, 3 255
          DA472065.1 - 2, 3 406
          DA894064.1 - 3, 4 294
          EU668324.1 - 3, 4 344
          KU178368.1 - 2, 3 276
          KU178369.1 - 2, 3 276
          KU178370.1 - 2, 3 276
          X04201.1 - 3, 4 445
          X04588.1 - 2, 3 328

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          actin binding motor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          movement of cell or subcellular component
          muscle contraction
          muscle filament sliding
          molecular_function
          actin binding
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0