siRNA Details
Gene Symbol: MT1E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005255956.4 | XP_005256013.1 | 2 | 396 | ||
GenBank mRNA | AF495759.1 | - | 2 | 282 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
