Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ITGB1BP1 SIRNA Assays ›
Gene Symbol:
ITGB1BP1
Gene Name:
integrin subunit beta 1 binding protein 1
Gene Aliases:
ICAP-1A, ICAP-1B, ICAP-1alpha, ICAP1, ICAP1A, ICAP1B
Chromosome Location:
Chr.2: 9405681 - 9423569 on Build GRCh38
UniGene ID:
Hs.467662
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
44218
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ITGB1BP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001319066.1
NP_001305995.1
4, 5
640
NM_001319067.1
NP_001305996.1
4, 5
542
NM_001319068.1
NP_001305997.1
4, 5
513
NM_001319069.1
NP_001305998.1
2, 3
342
NM_004763.4
NP_004754.1
4, 5
528
NM_022334.4
NP_071729.1
4, 5
528
NR_134951.1
4, 5
517
NR_134953.1
3, 4
421
NR_134954.1
4, 5
528
NR_134956.1
3, 4
449
NR_134957.1
3, 4
421
XM_005246183.4
XP_005246240.1
5, 6
1214
XM_005246184.4
XP_005246241.1
5, 6
653
XM_005246185.4
XP_005246242.1
5, 6
526
XM_005246189.4
XP_005246246.1
5, 6
1179
XM_006711903.3
XP_006711966.1
4, 5
758
XM_011510416.2
XP_011508718.1
5, 6
848
XM_017005267.1
XP_016860756.1
5, 6
2343
XM_017005268.1
XP_016860757.1
5, 6
518
XM_017005269.1
XP_016860758.1
4, 5
1069
XM_017005270.1
XP_016860759.1
5, 6
2352
GenBank mRNA
AF012023.1
-
4, 5
464
AF012024.1
-
4, 5
464
AI144519.1
-
4, 5
554
AI336197.1
-
4, 5
456
AK093179.1
-
4, 5
1102
AK289817.1
-
4, 5
441
AK299017.1
-
2, 3
235
BC012264.1
-
4, 5
420
3863540
BG032225.1
-
4, 5
421
BM823938.1
-
3, 4
248
BQ421551.1
-
4, 5
412
BQ425555.1
-
4, 5
473
BU520903.1
-
4, 5
487
BX647099.1
-
3, 4
381
CR457006.1
-
3, 4
296
CT002942.1
-
3, 4
329
DA751320.1
-
3, 4
305
DB167886.1
-
4, 5
423
DC321859.1
-
2, 3
229
DC361372.1
-
2, 3
345
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_008403, AK147929, AJ001373, AK004172, BC028772, and others.
Gene Ontology Categories:
Function(s)
Process(es)
blood vessel endothelial cell proliferation involved in sprouting angiogenesis
transcription, DNA-templated
negative regulation of protein kinase activity
negative regulation of cell adhesion involved in substrate-bound cell migration
cell-matrix adhesion
Notch signaling pathway
integrin-mediated signaling pathway
positive regulation of cell proliferation
negative regulation of cell proliferation
positive regulation of endothelial cell migration
negative regulation of fibroblast migration
protein transport
cell migration
cell differentiation
biomineral tissue development
negative regulation of protein binding
activation of protein kinase B activity
integrin activation
regulation of cell adhesion mediated by integrin
tube formation
intracellular signal transduction
cellular response to vascular endothelial growth factor stimulus
regulation of GTPase activity
receptor clustering
cellular response to fibroblast growth factor stimulus
positive regulation of Notch signaling pathway
positive regulation of transcription from RNA polymerase II promoter
regulation of blood vessel size
myoblast migration
positive regulation of stress fiber assembly
positive regulation of cell division
positive regulation of focal adhesion assembly
negative regulation of focal adhesion assembly
positive regulation of protein kinase B signaling
negative regulation of ERK1 and ERK2 cascade
negative regulation of cell migration involved in sprouting angiogenesis
positive regulation of protein targeting to membrane
negative regulation of protein targeting to membrane
negative regulation of substrate adhesion-dependent cell spreading
regulation of integrin-mediated signaling pathway
GDP-dissociation inhibitor activity
integrin binding
protein binding
protein transporter activity
protein kinase binding
protein complex binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-fz5f2:80/100.66.72.225:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0