Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001280796.1 | NP_001267725.1 | 1, 2 | 475 | ||
NM_001280797.1 | NP_001267726.1 | 1, 2 | 475 | |||
NM_013267.3 | NP_037399.2 | 1, 2 | 475 | |||
GenBank mRNA | AF110330.1 | - | 1, 2 | 230 | ||
AF223944.1 | - | 1, 2 | 402 | |||
AK000467.1 | - | 1, 2 | 222 | |||
AK289471.1 | - | 1, 2 | 179 | |||
AK294822.1 | - | 1, 2 | 420 | |||
AK303772.1 | - | 1, 2 | 310 | |||
BC025661.1 | - | 1, 2 | 190 | |||
BC119767.1 | - | 1, 2 | 420 | |||
BC119768.1 | - | 1, 2 | 420 |