Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TP53TG3C SIRNA Assays ›
          Gene Symbol:
          TP53TG3C LOC102723655 LOC102723713 TP53TG3 TP53TG3B TP53TG3D TP53TG3E TP53TG3F
          Gene Name:
          TP53 target 3C
          TP53-target gene 3 protein
          TP53-target gene 3 protein
          TP53 target 3
          TP53 target 3B
          TP53 target 3D
          TP53 target 3 family member E
          TP53 target 3 family member F
          Gene Aliases:
          TP53TG3, TP53TG3E, TP53TG3F
          -
          -
          P53TG3, TP53TG3A, TP53TG3E, TP53TG3F
          TP53TG3E, TP53TG3F
          TP53TG3, TP53TG3B, TP53TG3C, TP53TG3E, TP53TG3F
          -
          -
          Chromosome Location:
          Chr.16: 33191770 - 33196858 on Build GRCh38
          Chromosome Location:
          Chr.16|NT_187383: 14735 - 19833 on Build GRCh38
          Chromosome Location:
          Chr.16|NT_187383: 933861 - 936780 on Build GRCh38
          Chromosome Location:
          Chr.16: 32673528 - 32678621 on Build GRCh38
          Chromosome Location:
          Chr.16: 33358385 - 33363478 on Build GRCh38
          Chromosome Location:
          Chr.16: 32251926 - 32255928 on Build GRCh38
          Chromosome Location:
          Chr.16: 33301849 - 33306938 on Build GRCh38
          Chromosome Location:
          Chr.16: 33457155 - 33462249 on Build GRCh38
          UniGene ID:
          Hs.592038
          -
          -
          Hs.592038
          Hs.592038
          Hs.513537
          -
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID 44244

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TP53TG3C
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001205259.2 NP_001192188.1 1 209
          NR_110914.1 1 209
          XM_006721074.2 XP_006721137.1 1 185
          XM_011545920.1 XP_011544222.1 1 185
          XM_017023591.1 XP_016879080.1 1 2703
          XM_017023592.1 XP_016879081.1 1 192
          XM_017023593.1 XP_016879082.1 1 192
          GenBank mRNA DB053188.1 - 1 93
          DT215671.1 - 1 42
          Gene Symbol: LOC102723655
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_006724904.3 XP_006724967.1 1 2708
          XM_006724905.3 XP_006724968.1 1 2704
          XM_017030123.1 XP_016885612.1 1 2703
          XM_017030124.1 XP_016885613.1 1 2703
          Gene Symbol: LOC102723713
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_011546237.1 XP_011544539.1 1 523
          XM_011546238.1 XP_011544540.1 1 523
          XM_017030126.1 XP_016885615.1 1 523
          XM_017030127.1 XP_016885616.1 1 523
          Gene Symbol: TP53TG3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_016212.4 NP_057296.1 1 211
          NR_110886.1 1 211
          NR_110910.1 1 211
          XM_006721026.1 XP_006721089.1 1 185
          XM_011545779.1 XP_011544081.1 1 185
          XM_017023102.1 XP_016878591.1 1 2703
          XM_017023103.1 XP_016878592.1 1 192
          GenBank mRNA AB023508.1 - 1 211
          AK312220.1 - 1 192
          AL133588.1 - 1 814
          BC096733.1 - 1 209 40015196
          BC098165.1 - 1 209 40015195
          BX381390.2 - 1 103
          Gene Symbol: TP53TG3B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001099687.3 NP_001093157.1 1 211
          NR_110897.1 1 211
          NR_110911.1 1 211
          XM_006721084.1 XP_006721147.1 1 185
          XM_011545933.1 XP_011544235.1 1 185
          XM_017023638.1 XP_016879127.1 1 2703
          XM_017023639.1 XP_016879128.1 1 192
          Gene Symbol: TP53TG3D
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001243722.2 NP_001230651.1 1 209
          NR_110898.1 1 209
          XM_017023633.1 XP_016879122.1 1 1606
          XM_017023634.1 XP_016879123.1 1 1606
          XM_017023635.1 XP_016879124.1 1 1607
          XM_017023636.1 XP_016879125.1 1 1607
          XM_017023637.1 XP_016879126.1 1 1607
          GenBank mRNA AB023509.1 - 1 210
          BC096700.1 - 1 209 40015193
          BC099842.1 - 1 209 40015192
          CR541650.1 - 1 179
          DA077802.1 - 1 189
          Gene Symbol: TP53TG3E
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_006721117.2 XP_006721180.1 1 185
          XM_011546032.1 XP_011544334.1 1 185
          XM_017023950.1 XP_016879439.1 1 2703
          XM_017023951.1 XP_016879440.1 1 2703
          XM_017023952.1 XP_016879441.1 1 192
          XM_017023953.1 XP_016879442.1 1 192
          XM_017023954.1 XP_016879443.1 1 2704
          Gene Symbol: TP53TG3F
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_006721118.1 XP_006721181.1 1 185
          XM_011546033.1 XP_011544335.1 1 185
          XM_017023955.1 XP_016879444.1 1 2703
          XM_017023956.1 XP_016879445.1 1 2703
          XM_017023957.1 XP_016879446.1 1 192
          XM_017023958.1 XP_016879447.1 1 192
          XM_017023959.1 XP_016879448.1 1 2704

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0