Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WAC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016628.4 | NP_057712.2 | 3 | 221 | ||
NM_100264.2 | NP_567822.1 | 3 | 512 | |||
NM_100486.3 | NP_567823.1 | 3 | 221 | |||
XM_011519491.2 | XP_011517793.1 | 3 | 822 | |||
XM_017016315.1 | XP_016871804.1 | 3 | 332 | |||
XM_017016316.1 | XP_016871805.1 | 3 | 822 | |||
XM_017016317.1 | XP_016871806.1 | 3 | 332 | |||
XM_017016318.1 | XP_016871807.1 | 3 | 332 | |||
GenBank mRNA | AF116666.2 | - | 4 | 286 | ||
AK055852.1 | - | 3 | 329 | |||
AK223600.1 | - | 3 | 219 | |||
AK225846.1 | - | 3 | 325 | |||
AK290174.1 | - | 3 | 226 | |||
AK297460.1 | - | 3 | 134 | |||
AL713727.1 | - | 1 | 351 | |||
BC004258.1 | - | 3 | 212 | 3347345 | ||
BI226998.1 | - | 3 | 109 | |||
BI463901.1 | - | 3 | 612 | |||
CB143386.1 | - | 3 | 400 |